Skip to Content
Merck
All Photos(2)

Key Documents

SAB1410166

Sigma-Aldrich

Anti-NUDT5 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

YSA1, YSA1H, hYSAH1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 24.3 kDa

species reactivity

human

technique(s)

immunofluorescence: suitable
western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NUDT5(11164)

General description

Nudix hydrolases, such as NUDT5, eliminate toxic nucleotide derivatives from the cell and regulate the levels of important signaling nucleotides and their metabolites (McLennan, 1999 [PubMed 10373642]).[supplied by OMIM

Immunogen

NUDT5 (NP_054861.2, 1 a.a. ~ 219 a.a) full-length human protein.

Sequence
MESQEPTESSQNGKQYIISEELISEGKWVKLEKTTYMDPTGKTRTWESVKRTTRKEQTADGVAVIPVLQRTLHYECIVLVKQFRPPMGGYCIEFPAGLIDDGETPEAAALRELEEETGYKGDIAECSPAVCMDPGLSNCTIHIVTVTINGDDAENARPKPKPGDGEFVEVISLPKNDLLQRLDALVAEEHLTVDARVYSYALALKHANAKPFEVPFLKF

Biochem/physiol Actions

NUDT5 (Nudix, nucleoside diphosphate linked moiety X-type motif 5) is mainly involved in maintaining the intracellular level of ADP-ribose by hydrolyzing ADP-ribose (ADPR) to AMP and ribose 5′-phosphate. It also restricts non-enzymatic ADP-ribosylation. Its N-terminal antiparallel β-sheet plays an important role in dimerization, which is essential for substrate recognition and enzymatic activity. NUDT5 has also been predicted for altered cell proliferation. It has been suggested that cells with suppressed NUDT5 shows delayed cell cycle G1 phase. Thus, it has been concluded that NUDT5 may involve in cell growth and cell cycle proliferation by regulating G1-S transition in mammalian cells.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Manwu Zha et al.
Journal of molecular biology, 364(5), 1021-1033 (2006-10-21)
Human NUDT5 (hNUDT5) is an ADP-ribose pyrophosphatase (ADPRase) belonging to the Nudix hydrolase superfamily. It presumably plays important roles in controlling the intracellular level of ADP-ribose (ADPR) to prevent non-enzymatic ADP-ribosylation by hydrolyzing ADPR to AMP and ribose 5'-phosphate. We
Hong-Nu Yu et al.
Biochemical and biophysical research communications, 354(3), 764-768 (2007-01-31)
The ADP-ribose (ADPR) pyrophosphatase (ADPRase) NUDT5, a member of a superfamily of Nudix hydrolases, hydrolyzes ADP-ribose (ADPR) to AMP and ribose 5'-phosphate. Nitric oxide (NO) enhances nonenzymatic ADP-ribosylation of proteins such as beta-actin and glyceraldehydes 3-phosphate dehydrogenase in the presence
Li-Qun Zhang et al.
Molecular and cellular biochemistry, 363(1-2), 377-384 (2011-12-28)
The molecule 8-oxo-7,8-dihydroguanine (8-oxoGua), an oxidized form of guanine, can pair with adenine or cytosine during nucleic acid synthesis. Moreover, RNA containing 8-oxoGua causes translational errors, thus leading to the production of abnormal proteins. Human NUDT5, a MutT-related protein, catalyzes

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service