Skip to Content
Merck
All Photos(1)

Key Documents

SAB1408588

Sigma-Aldrich

Anti-IFI27 antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

FAM14D, ISG12, ISG12A, P27

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~11.3 kDa

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IFI27(3429)

Immunogen

IFI27 (NP_005523.3, 1 a.a. ~ 119 a.a) full-length human protein.

Sequence
MEASALTSSAVTSVAKVVRVASGSAVVLPLARIATVVIGGVVAVPMVLSAMGFTAAGIASSSIAAKMMSAAAIANGGGVASGSLVATLQSLGATGLSGLTKFILGSIGSAIAAVIARFY

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tuoyang Li et al.
International journal of molecular sciences, 23(19) (2022-10-15)
A disintegrin and metalloproteinase with thrombospondin motifs 16 (ADAMTS16) has been reported to be involved in the pathogenesis of solid cancers. However, its role in gastric cancer (GC) is unclear. In this study, the role of ADAMTS16 in gastric cancer
Rilin Deng et al.
Cellular & molecular immunology, 17(11), 1163-1179 (2020-09-24)
The ability to harness innate immunity is a promising solution for improving cancer immunotherapy. Interferon (IFN) induces expression of IFN-stimulated genes (ISGs) by activating the JAK-STAT signaling pathway to promote innate immunity and inhibit malignant tumor growth, but the functions
Rilin Deng et al.
iScience, 27(4), 109533-109533 (2024-04-09)
Hepatitis B virus (HBV) infection generally elicits weak type-I interferon (IFN) immune response in hepatocytes, covering the regulatory effect of IFN-stimulated genes. In this study, low level of IFN-stimulated gene 12a (ISG12a) predicted malignant transformation and poor prognosis of HBV-associated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service