Skip to Content
Merck
All Photos(2)

Key Documents

SAB1403744

Sigma-Aldrich

Monoclonal Anti-DRD2, (N-terminal) antibody produced in mouse

clone 1B11, ascites fluid

Synonym(s):

D2DR, D2R

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1B11, monoclonal

mol wt

antigen ~38.1 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1:500-1:1000

isotype

IgG2aκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... DRD2(1813)

General description

This gene encodes the D2 subtype of the dopamine receptor. This G-protein coupled receptor inhibits adenylyl cyclase activity. A missense mutation in this gene causes myoclonus dystonia; other mutations have been associated with schizophrenia. Alternative splicing of this gene results in two transcript variants encoding different isoforms. A third variant has been described, but it has not been determined whether this form is normal or due to aberrant splicing. (provided by RefSeq)

Immunogen

DRD2 (AAH21195, 1 a.a. ~ 110 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MDPLNLSWYDDDLERQNWSRPFNGSDGKADRPHYNYYATLLTLLIAVIVFGNVLVCMAVSREKALQTTTNYLIVSLAVADLLVATLVMPWVVYLEVVGEWKFSRIHCDIF

Physical form

Clear solution

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mei Zhou et al.
Neurotoxicity research, 36(2), 376-386 (2019-06-16)
Repeated methamphetamine (METH) exposure can cause severe neurotoxicity to the central nervous system, and lead to memory deficits. L-Stepholidine (L-SPD) is a structurally identified alkaloid extract of the Chinese herb Stephania intermedia, which elicits dopamine (DA) D1-type receptors partial agonistic
A N DeCarlo et al.
Animal reproduction science, 207, 73-82 (2019-06-19)
A dopamine type-2 receptor (DRD2) SNP, previously found to be correlated with serum prolactin (PRL) concentrations in cattle, was evaluated for impact on growth traits, serum prolactin concentration, and semen quality. Over a four-year period, yearling beef bulls were allowed
Rebecca D Pratt et al.
Scientific reports, 9(1), 7927-7927 (2019-05-30)
Obesity has become a worldwide epidemic. We have previously reported that systemic administration of pNaKtide which targets the Na/K-ATPase oxidant amplification loop (NKAL) was able to decrease systemic oxidative stress and adiposity in mice fed a high fat and fructose
Rafael L Batista et al.
American journal of clinical oncology, 42(2), 221-227 (2018-12-13)
Complete tumor removal by transsphenoidal surgery is usually difficult for large nonfunctioning pituitary adenomas (NFPAs). A validated medical treatment may be useful for their management. This study evaluates the clinical efficacy of the dopaminergic agonist cabergoline for residual NFPA. We
Koji Ohira
Biochemical and biophysical research communications, 512(3), 577-583 (2019-03-27)
Cortical GABAergic interneurons originate and migrate tangentially from the medial ganglionic eminence (MGE), but its mechanism remains unknown. In this study, we show that dopamine (DA) stimulates the differentiation and migration of cortical interneurons derived from MGE cells. Using immunohistochemistry

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service