Skip to Content
Merck
All Photos(1)

Key Documents

SAB1403066

Sigma-Aldrich

Monoclonal Anti-CDIPT antibody produced in mouse

clone 1F8, ascites fluid

Synonym(s):

MGC1328, PIS, PIS1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

primary antibodies

clone

1F8, monoclonal

mol wt

antigen ~49.54 kDa

species reactivity

human

technique(s)

indirect ELISA: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDIPT(10423)

Related Categories

General description

Phosphatidylinositol breakdown products are ubiquitous second messengers that function downstream of many G protein-coupled receptors and tyrosine kinases regulating cell growth, calcium metabolism, and protein kinase C activity. Two enzymes, CDP-diacylglycerol synthase and phosphatidylinositol synthase, are involved in the biosynthesis of phosphatidylinositol. Phosphatidylinositol synthase, a member of the CDP-alcohol phosphatidyl transferase class-I family, is an integral membrane protein found on the cytoplasmic side of the endoplasmic reticulum and the Golgi apparatus. (provided by RefSeq)

Immunogen

CDIPT (AAH01444, 1 a.a. ~ 213 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MPDENIFLFVPNLIGYARIVFAIISFYFMPCCPLTASSFYLLSGLLDAFDGHAARALNQGTRFGAMLDMLTDRCSTMCLLVNLALLYPGATLFFQISMSLDVASHWLHLHSSVVRGSESHKMIDLSGNPVLRIYYTSRPALFTLCAGNELFYCLLYLFHFSEGPLVGSVGLFRMGLWVTAPIALLKSLISVIHLITAARNMAALDAADRAKKK

Physical form

Clear solution

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service