Skip to Content
Merck
All Photos(2)

Key Documents

SAB1402973

Sigma-Aldrich

Monoclonal Anti-PXDN antibody produced in mouse

clone 2C11, purified immunoglobulin, buffered aqueous solution

Synonym(s):

D2S448, D2S448E, KIAA0230, MG50, PRG2, PXN, VPO

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

2C11, monoclonal

form

buffered aqueous solution

mol wt

antigen ~38.21 kDa

species reactivity

human

technique(s)

capture ELISA: suitable
indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PXDN(7837)

General description

Drosophila peroxidasin is an extracellular matrix-associated peroxidase (Horikoshi et al., 1999 [PubMed 10441517]). It is expressed exclusively in hemocytes derived from head mesoderm at a very early stage of differentiation. Peroxidasin exists as a homotrimer with a unique hybrid structure that combines an enzymatically functional peroxidase domain with motifs that are typically found in extracellular matrix-associated proteins. It is a secreted protein that contains a secretory recognition sequence at its N terminus. Peroxidasin catalyzes hydrogen peroxide-driven radioiodination, oxidations, and the formation of dityrosine in vitro. It is also thought to function in extracellular matrix consolidation, phagocytosis, and defense.[supplied by OMIM] Peroxidasin (PXDN) is a multidomain, glycosylated, homotrimeric peroxidase. It is expressed in the endoplasmic reticulum and secreted into the extracellular matrix. The 1479-amino acid protein belongs to the peroxidase-cyclooxygenase superfamily. It possesses a catalytic peroxidase domain (POX), a leucine-rich repeat domain (LRR), four C-like immunoglobulin domains (Ig) at the amino-terminal and a carboxy-terminal von Willebrand factor type C module (VWC). The gene encoding PXDN has 23 exons and is localized on human chromosome 2p25.3.

Immunogen

PXDN (XP_056455, 1452 a.a. ~ 1561 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
STSAFSTRSDASGTNDFREFVLEMQKTITDLRTQIKKLESRLSTTECVDAGGESHANNTKWKKDACTICECKDGQVTCFVEACPPATCAVPVNIPGACCPVCLQKRAEEK

Biochem/physiol Actions

Peroxidasin (PXDN) releases hypobromous acid, which is involved in collagen IV reinforcement. The protein stabilizes the basement membrane by enhancing the covalent crosslinks in the collagen IV network. Mutations in the gene encoding PXDN have been linked to microphthalmia and anterior segment dysgenesis.

Physical form

Solution in phosphate buffered saline, pH 7.4

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selene Colon et al.
American journal of physiology. Renal physiology, 316(2), F360-F371 (2018-12-20)
Renal fibrosis is the pathological hallmark of chronic kidney disease (CKD) and manifests as glomerulosclerosis and tubulointerstitial fibrosis. Reactive oxygen species contribute significantly to renal inflammation and fibrosis, but most research has focused on superoxide and hydrogen peroxide (H2O2). The
Novel mutations in PXDN cause microphthalmia and anterior segment dysgenesis.
Choi A
European Journal of Human Genetics (2015)
Pre-steady-state Kinetics Reveal the Substrate Specificity and Mechanism of Halide Oxidation of Truncated Human Peroxidasin 1.
Paumann-Page M
The Journal of Biological Chemistry (2017)
Jodi Dougan et al.
International journal of molecular sciences, 20(12) (2019-06-27)
Peroxidasin (PXDN), a human homolog of Drosophila PXDN, belongs to the family of heme peroxidases and has been found to promote oxidative stress in cardiovascular tissue, however, its role in prostate cancer has not been previously elucidated. We hypothesized that
Microduplications Disrupting the MYT1L Gene (2p25.3) are Associated with Schizophrenia
Yohan Lee
Psychiatric Genetics (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service