Skip to Content
Merck
All Photos(1)

Key Documents

SAB1401025

Sigma-Aldrich

Anti-ATP6V0B antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym(s):

ATP6F, HATPL, VMA16

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

western blot: 1 μg/mL

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ATP6V0B(533)

Related Categories

General description

This gene encodes a component of vacuolar ATPase (V-ATPase), a multisubunit enzyme that mediates acidification of eukaryotic intracellular organelles. V-ATPase dependent organelle acidification is necessary for such intracellular processes as protein sorting, zymogen activation, receptor-mediated endocytosis, and synaptic vesicle proton gradient generation. V-ATPase is composed of a cytosolic V1 domain and a transmembrane V0 domain. The V1 domain consists of three A and three B subunits, two G subunits plus the C, D, E, F, and H subunits. The V1 domain contains the ATP catalytic site. The V0 domain consists of five different subunits: a, c, c′, c′′, and d. Additional isoforms of many of the V1 and V0 subunit proteins are encoded by multiple genes or alternatively spliced transcript variants. This encoded protein is part of the transmembrane V0 domain and is the human counterpart of yeast VMA16. Two alternatively spliced transcript variants that encode different proteins have been found for this gene. (provided by RefSeq)

Immunogen

ATP6V0B (NP_004038.1, 1 a.a. ~ 205 a.a) full-length human protein.

Sequence
MTGLALLYSGVFVAFWACALAVGVCYTIFDLGFRFDVAWFLTETSPFMWSNLGIGLAISLSVVGAAWGIYITGSSIIGGGVKAPRIKTKNLVSIIFCEAVAIYGIIMAIVISNMAEPFSATDPKAIGHRNYHAGYSMFGAGLTVGLSNLFCGVCVGIVGSGAALADAQNPSLFVKILIVEIFGSAIGLFGVIVAILQTSRVKMGD

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Solution in phosphate buffered saline, pH 7.4

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service