Skip to Content
Merck
All Photos(1)

Key Documents

SAB1306537

Sigma-Aldrich

ANTI-BECLIN1-BH3 DOMAIN antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

BECN1, Beclin-1, Coiled-coil myosin-like BCL2-interacting protein, GT197, Protein GT197

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

51896 Da

species reactivity

human, mouse

technique(s)

flow cytometry: 1:10-1:50
immunohistochemistry: 1:50-1:100
western blot: 1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... BECN1(8678)

General description

Beclin 1 (BECN1) is a tumor suppressor gene. It is a 450 amino-acid protein having three domains. The gene encoding BECN1 is localized on human chromosome 17q21.

Application

ANTI-BECLIN1-BH3 DOMAIN antibody produced in rabbit has been used in Western blotting.

Biochem/physiol Actions

Beclin 1 (BECN1) acts as an initiator of autophagy and is vital in the early stages of autophagosome formation. It has a role in various tumors and cancers. BECN1 is a component of the phosphatidylinositol-3-kinase (PI3K) complex which mediates vesicle-trafficking.

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Class III phosphoinositide 3-kinase?Beclin1 complex mediates the amino
acid-dependent regulation of autophagy in C2C12 myotubes
BioChemistry: An Indian Journal (2003)
Polyglutamine tracts regulate beclin 1-dependent autophagy.
Ashkenazi A, et al.
Nature, 545, 108-111 (2017)
RNF146 Inhibits Excessive Autophagy by Modulating the Wnt-?-Catenin Pathway in Glutamate Excitotoxicity Injury
Yuefan Yang
Frontiers in Cellular Neuroscience (2017)
Decreased expression of Beclin?1 is significantly associated with a poor prognosis in oral tongue squamous cell carcinoma.
Hu Z
Molecular Medicine Reports, 14(2), 1567-1573 (2016)
Decreased expression of Beclin-1 is significantly associated with a poor prognosis in oral tongue squamous cell carcinoma.
Hu Z, et.al
Molecular Medicine Reports, 14(2), 1567-1573 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service