Skip to Content
Merck
All Photos(1)

Key Documents

SAB1305554

Sigma-Aldrich

Anti-β-Actin (ACTB) Antibody

mouse monoclonal, 137CT26.1.1

Synonym(s):

ACTB, Actin, cytoplasmic 1, Actin, cytoplasmic 1, N-terminally processed, Beta-actin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

Product Name

MONOCLONAL ANTI-BETA-ACTIN antibody produced in mouse, clone 137CT26.1.1, IgG fraction of antiserum, buffered aqueous solution

biological source

mouse

Quality Level

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

137CT26.1.1, monoclonal

form

buffered aqueous solution

mol wt

41737 Da

species reactivity

mouse, human

technique(s)

immunofluorescence: 1:10-1:50
immunohistochemistry: 1:25
western blot: 1:1000

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ACTB(60)

General description

β-actin (ACTB), a member of the actin multigene family, is an isoform of actin-associated with non-muscle cells. ACTB is a housekeeping gene mapped to human chromosome 7p22.1.

Application

Monoclonal Anti-Beta-Actin antibody produced in mouse has been used in western blotting at a dilution of 1:1000.

Biochem/physiol Actions

β-actin (ACTB) forms the core of the cell cytoskeleton. The polymerization of β-actin is essential for cell physiology. It may serve as a potential chemotherapeutic agent. Mutation in the ACTB gene is implicated in sensory hearing loss, delayed-onset generalized dystonia and a combination of ventral midline malformations. A loss-of-function mutation in the ACTB gene may cause intellectual disability.

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Induction of drug resistance in human hepatoma cells cultured on a silicate fiber based 3d scaffold
Mizutami T, et al.
Advances in Biochemical Engineering null
Sara Cuvertino et al.
American journal of human genetics, 101(6), 1021-1033 (2017-12-09)
ACTB encodes β-actin, an abundant cytoskeletal housekeeping protein. In humans, postulated gain-of-function missense mutations cause Baraitser-Winter syndrome (BRWS), characterized by intellectual disability, cortical malformations, coloboma, sensorineural deafness, and typical facial features. To date, the consequences of loss-of-function ACTB mutations have
Ioannis Panagopoulos et al.
Anticancer research, 40(3), 1239-1245 (2020-03-07)
Since the first description of five pericytomas with the t(7;12)/ACTB-GLI1 fusion gene, only three new tumors were studied by both cytogenetics and molecular techniques. We report here genetic data on another case of this rare tumor. Cytogenetic, fluorescence in situ
Immacolata Andolfo et al.
Blood advances, 7(12), 2681-2693 (2023-01-04)
Gain-of-function mutations in PIEZO1 cause dehydrated hereditary stomatocytosis (DHS) or hereditary xerocytosis, an autosomal dominant hemolytic anemia characterized by high reticulocyte count, a tendency to macrocytosis, and mild jaundice, as well as by other variably penetrant clinical features, such as
Sequence and structural analysis of beta-actin protein of fishes, using bioinformatics tools and techniques
Acharya UR, et al.
International journal of biosciences, 4(11) (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service