Skip to Content
Merck
All Photos(1)

Key Documents

SAB1303565

Sigma-Aldrich

ANTI-ADAMTS13 (CENTER) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym(s):

A disintegrin and metalloproteinase with thrombospondin motifs 13, ADAM-TS 13, ADAM-TS13, ADAMTS-13, ADAMTS13, C9orf8, vWF-CP, vWF-cleaving protease, von Willebrand factor-cleaving protease

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

153604 Da

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:100
western blot: 1:250-1:500

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

A disintegrin-like and metalloprotease with thrombospondin type 1 motif, 13 (ADAMTS13) also known as von willebrand factor-cleaving protease (VWFCP), has distinct regions like a metalloproteinase domain, a disintegrin-like domain, and a thrombospondin type 1 domain and spacer domain rich in cysteine residues. ADAMST13 is highly expressed in the liver and secreted as an active enzyme in the blood. In human chromosome, the gene ADAMTS13 is localised on 9q34.

Biochem/physiol Actions

A disintegrin-like and metalloprotease with thrombospondin type 13 (ADAMTS13) cleaves von Willebrand factor (VWF) and thereby controls platelet thrombus formation. ADAMTS13 requires zinc, and calcium for its effective catalytic activity. Mutations in ADAMTS13 can lead to life threatening acquired thrombotic thrombocytopenic purpura (TTP) or hemolytic-uremic syndrome. Congenital deficiency of ADAMTS13 leads to Upshaw−Schulman syndrome.

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide. This antibody is purified through a protein A column, followed by peptide affinity purification.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mutations in a member of the ADAMTS gene family cause thrombotic thrombocytopenic purpura
Levy GG, et al.
Nature, 413(6855), 488-488 (2001)
Crystal structures of the noncatalytic domains of ADAMTS13 reveal multiple discontinuous exosites for von Willebrand factor
Akiyama M, et al.
Proceedings of the National Academy of Sciences of the USA, 106(46), 19274-19279 (2009)
Novel compound heterozygote mutations (H234Q/R1206X) of the ADAMTS13 gene in an adult patient with Upshaw-Schulman syndrome showing predominant episodes of repeated acute renal failure
Shibagaki Y, et al.
Nephrology, Dialysis, and Transplantation, 21(5), 1289-1292 (2006)
A functional calcium-binding site in the metalloprotease domain of ADAMTS13
Gardner MD, et al.
Blood, 113(5), 1149-1157 (2009)
Zinc and calcium ions cooperatively modulate ADAMTS13 activity
Anderson PJ, et al.
The Journal of Biological Chemistry, 281(2), 850-857 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service