Skip to Content
Merck
All Photos(1)

Key Documents

S8195

Sigma-Aldrich

Swainsonine

from Metarrhizium anisopliae, ≥98% (TLC)

Synonym(s):

(1S,2R,8R,8aR)-1,2,8-Octahydroindolizidinetriol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H15NO3
CAS Number:
Molecular Weight:
173.21
Beilstein:
4175740
MDL number:
UNSPSC Code:
51102829
PubChem Substance ID:
NACRES:
NA.85
Pricing and availability is not currently available.

biological source

Metarrhizium anisopliae

Quality Level

Assay

≥98% (TLC)

form

lyophilized powder

storage condition

(Keep container tightly closed in a dry and well-ventilated place.)

color

white to faint yellow

solubility

H2O: soluble 1 mg/mL

antibiotic activity spectrum

neoplastics

Mode of action

enzyme | inhibits

storage temp.

2-8°C

SMILES string

O[C@@H]1CCCN2C[C@@H](O)[C@@H](O)C12

InChI

1S/C8H15NO3/c10-5-2-1-3-9-4-6(11)8(12)7(5)9/h5-8,10-12H,1-4H2/t5-,6-,7?,8-/m1/s1

InChI key

FXUAIOOAOAVCGD-DCDLSZRSSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Swainsonine is a potent inhibitor of various α-mannosidases, especially of α-mannosidase II. It inhibits glycoprotein processing and also acts as immune modulator.
Swainsonine is an indolizidine alkaloid from the plant Metarrhizium anisopliae that is used as a potent α-mannosidase inhibitor[1]. Product S8195 has been used in chemical inhibition assays of CHO Lec2 cells to inhibit glycosylation [2].

Biochem/physiol Actions

Swainsonine is a potent α-mannosidase inhibitor. It also has antimetastatic, antiproliferative, and immunomodulatory activity [1]. It also inhibits glycoprotein processing.

Packaging

1MG

Preparation Note

Soluble in water, methanol, DMSO

Other Notes

Keep container tightly closed in a dry and well-ventilated place.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shen Shen et al.
The Journal of biological chemistry, 286(15), 13532-13540 (2011-02-19)
Sialylated glycans serve as cell surface attachment factors for a broad range of pathogens. We report an atypical example, where desialylation increases cell surface binding and infectivity of adeno-associated virus (AAV) serotype 9, a human parvovirus isolate. Enzymatic removal of
Julien Louvel et al.
Organic letters, 13(24), 6452-6455 (2011-11-16)
The asymmetric synthesis of (-)-swainsonine and (-)-8-epi-swainsonine is reported through the addition of either the allenylzinc or the allenyl lithio cyanocuprate reagents derived from [3-(methoxymethoxy)prop-1-ynyl]trimethylsilane to enantiopure α,β-dialkoxy N-tert-butanesulfinylimines derived from d-erythronolactone.
Fábio S Mendonça et al.
Journal of veterinary diagnostic investigation : official publication of the American Association of Veterinary Laboratory Diagnosticians, Inc, 24(1), 90-95 (2012-03-01)
A disease of the nervous system is reported in goats in the semiarid region of northeastern Brazil. Histological examination showed diffuse vacuolation of neurons and epithelial cells of the pancreas, thyroid, renal tubules, and liver. The swainsonine-containing plant Ipomoea verbascoidea
Guo-Dong Yang et al.
Toxicon : official journal of the International Society on Toxinology, 60(1), 44-49 (2012-04-10)
Endophytic Undifilum oxytropis found within toxic locoweeds (Astragalus and Oxytropis spp.) produces the indolizidine alkaloid swainsonine, which is responsible for locoism in grazing animals. The aim of the current study is to establish an easy and accurate method for the
Zhaocai Li et al.
International journal of biological sciences, 8(3), 394-405 (2012-03-07)
Swainsonine (1, 2, 8-trihyroxyindolizidine, SW), a natural alkaloid, has been reported to exhibit anti-cancer activity on several mouse models of human cancer and human cancers in vivo. However, the mechanisms of SW-mediated tumor regression are not clear. In this study

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service