Skip to Content
Merck
All Photos(1)

Key Documents

S7651

Sigma-Aldrich

Sphingomyelinase from Bacillus cereus

lyophilized powder, ≥100 units/mg protein

Synonym(s):

Sphingomyelin choline phosphohydrolase, Sphingomyelin phosphodiesterase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

lyophilized powder

Quality Level

specific activity

≥100 units/mg protein

mol wt

36873 Da

composition

Protein, ~10% Lowry

storage temp.

2-8°C

Gene Information

Bacillus cereus ATCC 14579 ... BC0671(1203022)

Looking for similar products? Visit Product Comparison Guide

General description

Sphingomyelinase from Bacillus cereus (Bc-Smase) is a Mg2+-containing metalloenzyme. Bacillus cereus sphingomyelinase induces hemolysis of sheep erythrocytes.
Sphingomyelinase modulates adenosine triphosphate (ATP)-independent endocytosis of fibroblast and large vesicles of macrophages. Sphingomyelinase causes hemolysis of human erythrocytes. It is also a potent virulence factor leading to invasive diseases.

Application

Sphingomyelinase from Bacillus cereus has been used:
  • in the measurement of membrane capacitance in brain slices
  • for testing and confirming its silkworm-killing activity
  • in enzyme activity assay and for elucidating its effect on hydrolysis of sphingomyelin in complex lipid mixtures

Sphingomyelinase has been used in a study to investigate skeletal manifestations in pediatric and adult patients with Niemann Pick disease.

Biochem/physiol Actions

Initiates the formation of sphingomyelin-based second messengers. Activates MAPK (mitogen-activated protein kinase) and SAPKs (stress-activated protein kinases); generates ceramide from sphingomyelin.

Unit Definition

One unit will hydrolyze 1.0 μmole of TNPAL-sphingomyelin per min at pH 7.4 at 37 °C.

Physical form

Lyophilized powder containing potassium phosphate buffer salts and stabilizer.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Masataka Oda et al.
Biochimica et biophysica acta, 1798(6), 1073-1080 (2010-03-11)
Bacillus cereus sphingomyelinase (Bc-SMase) induces hemolysis of sheep erythrocytes which contain large amounts of sphingomyelin. We investigated the mechanism of this hemolysis in comparison to that induced by Clostridium perfringens alpha-toxin. Pertussis toxin, a Gi-specific inhibitor, N-oleoylethernolamine, a ceramidase inhibitor
Purification of a soil bacteria exotoxin using silkworm toxicity to measure specific activity
Usui K, et al.
Microbial Pathogenesis, 46(2), 59-62 (2009)
Melissa Wasserstein et al.
Journal of inherited metabolic disease, 36(1), 123-127 (2012-06-22)
Niemann-Pick disease (NPD) due to acid sphingomyelinase deficiency is a lipid storage disease resulting from the accumulation of sphingomyelin, predominantly within cells of the monocyte-macrophage system. In contrast to other lysosomal storage disorders, skeletal involvement in NPD has not been
Lipid bilayers containing sphingomyelins and ceramides of varying N-acyl lengths: a glimpse into sphingolipid complexity
Jimenez-Rojo N, et al.
Biochimica et Biophysica Acta - Biomembranes, 1838(1), 456-464 (2014)
Masataka Oda
Yakugaku zasshi : Journal of the Pharmaceutical Society of Japan, 129(10), 1233-1238 (2009-10-03)
Bacillus cereus is one that causes of opportunistic human infections. Sphingomyelinase produced by B. cereus is assumed a virulence factor for the infection. Sphingomyelinase from Bacillus cereus (Bc-SMase) is Mg(2+)-containing metalloenzyme. Bc-SMase is a family of neutral SMase (nSMase) and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service