Skip to Content
Merck
All Photos(1)

Key Documents

S6942

Sigma-Aldrich

Staurosporine

from Streptomyces sp., >98% (HPLC), solution, protein kinase inhibitor

Synonym(s):

Antibiotic AM-2282

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77
Pricing and availability is not currently available.

Product Name

Staurosporine solution from Streptomyces sp., Ready Made Solution, 1 mM in DMSO (100 μg/214 μL), 0.2 μm filtered

Quality Level

sterility

0.2 μm filtered

Assay

>98% (HPLC)

concentration

1 mM in DMSO (100 μg/214 μL)

technique(s)

cell culture | mammalian: suitable

antibiotic activity spectrum

fungi

Mode of action

enzyme | inhibits

shipped in

wet ice

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: indol derivative

Application

Staurosporine solution from Streptomyces sp. was used to study the effect of PKC inhibition on signaling mediated by angiotensin II.3 It was used to induce cell death in Jurkat cells.4

Biochem/physiol Actions

Potent cell-permeable inhibitor of protein kinase C. Induces apoptosis in Jurkat cells.
Potent inhibitor of phospholipid/calcium-dependent protein kinase. Inhibits the upregulation of VEGF expression in tumor cells.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

188.6 °F - closed cup

Flash Point(C)

87 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dale W Hailey et al.
Cell, 141(4), 656-667 (2010-05-19)
Starvation-induced autophagosomes engulf cytosol and/or organelles and deliver them to lysosomes for degradation, thereby resupplying depleted nutrients. Despite advances in understanding the molecular basis of this process, the membrane origin of autophagosomes remains unclear. Here, we demonstrate that, in starved
Maren de Vries et al.
Journal of virology (2021-02-25)
Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2) is the etiological agent of Coronavirus Disease 2019 (COVID-19). There is a dire need for novel effective antivirals to treat COVID-19, as the only approved direct-acting antiviral to date is remdesivir, targeting the
Yu-Jung Lee et al.
American journal of physiology. Renal physiology, 292(1), F340-F350 (2006-08-10)
Vasopressin and angiotensin II (ANG II) play a major role in renal water and Na(+) reabsorption. We previously demonstrated that ANG II AT(1) receptor blockade decreases dDAVP-induced water reabsorption and AQP2 levels in rats, suggesting cross talk between these two
Jan Roger Olsen et al.
PloS one, 8(5), e62547-e62547 (2013-05-10)
The transcription factor p63 is central for epithelial homeostasis and development. In our model of epithelial to mesenchymal transition (EMT) in human prostate cells, p63 was one of the most down-regulated transcription factors during EMT. We therefore investigated the role
Suainibhe Kelly et al.
Nanoscale, 13(41), 17615-17628 (2021-10-19)
The use of nanomaterials as therapeutic delivery vehicles requires their careful pre-clinical evaluation. Of particular importance in this regard is measurement of cellular toxicity, ideally assessing multiple parameters in parallel from various relevant subcellular organelles. In recent years it has

Articles

Protein-based drug transporters are expressed in Sf9 cells. Understanding the specific mechanisms of tumor cell transporters is an essential aspect of chemotherapeutic drug design.

Protein-based drug transporters are expressed in Sf9 cells. Understanding the specific mechanisms of tumor cell transporters is an essential aspect of chemotherapeutic drug design.

Protein-based drug transporters are expressed in Sf9 cells. Understanding the specific mechanisms of tumor cell transporters is an essential aspect of chemotherapeutic drug design.

Protein-based drug transporters are expressed in Sf9 cells. Understanding the specific mechanisms of tumor cell transporters is an essential aspect of chemotherapeutic drug design.

See All

Questions

1–2 of 2 Questions  
  1. What concentration of Staurosporin is used for the release of microparticles from apoptotic cells?

    1 answer
    1. Staurosporin can induce apopotosis in Jurkat cells (106/ml) at 1 ug/ml following a 1-2 hour incubation at 37°C. For the release of microparticles from apoptotic Jurkat cells, use staurosporin at 1 uM for 107 cells/ml. Please see the reference below for further details. Apoptosis 15(5):586, 2010.

      Helpful?

  2. What is the Department of Transportation shipping information for this product?

    1 answer
    1. Transportation information can be found in Section 14 of the product's (M)SDS.To access the shipping information for this material, use the link on the product detail page for the product.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service