Skip to Content
Merck
All Photos(1)

Key Documents

S6448

Sigma-Aldrich

SYK, active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

79-107 nmol/min·mg

mol wt

~100 kDa

UniProt accession no.

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... SYK(6850)

Biochem/physiol Actions

SYK is a non-receptor protein tyrosine kinase that is widely expressed in hematopoietic cells. It is involved in coupling activated immunoreceptors to downstream signaling events that mediate diverse cellular responses, including proliferation, differentiation, and phagocytosis. In B cells, SYK plays a crucial role in intracellular signal transduction induced by oxidative stress as well as antigen receptor engagement.SYK has been shown to act as a potential tumor suppressor in breast cancer. Absence of SYK protein in primary breast tumors is correlated with poor outcomes. SYK deficient cells have increased motility that is restored to normalcy by replacement with wild-type SYK.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomoko Takano et al.
Antioxidants & redox signaling, 4(3), 533-541 (2002-09-07)
Oxidative stress induces the activation of multiple signaling pathways related to various cellular responses. In B cells, Syk has a crucial role in intracellular signal transduction induced by oxidative stress as well as antigen receptor engagement. Treatment of B cells
Yu Yu et al.
Oncogene, 37(28), 3778-3789 (2018-04-13)
Cell motility and invasiveness are prerequisites for dissemination, and largely account for cancer mortality. We have identified an actionable kinase, spleen tyrosine kinase (SYK), which is keenly tightly associated with tumor progression in ovarian cancer. Here, we report that active
Christopher S Navara
Current pharmaceutical design, 10(15), 1739-1744 (2004-06-08)
The spleen tyrosine kinase Syk is an enigmatic protein tyrosine kinase functional in a number of diverse cellular processes. It is best known as a non receptor protein tyrosine kinase involved in signal transduction in cells of hematopoietic origin and
Yu Yu et al.
EBioMedicine, 47, 184-194 (2019-09-08)
Spleen tyrosine kinase (SYK) is frequently upregulated in recurrent ovarian carcinomas, for which effective therapy is urgently needed. SYK phosphorylates several substrates, but their translational implications remain unclear. Here, we show that SYK interacts with EGFR and ERBB2, and directly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service