S5576
SKA-31
≥98% (HPLC)
Synonym(s):
Naphtho[1,2-d]thiazol-2-ylamine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C11H8N2S
CAS Number:
Molecular Weight:
200.26
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.25
Recommended Products
Quality Level
Assay
≥98% (HPLC)
form
solid
solubility
DMSO: ≥30 mg/mL
storage temp.
−20°C
SMILES string
Nc1nc2c(ccc3ccccc23)s1
InChI
1S/C11H8N2S/c12-11-13-10-8-4-2-1-3-7(8)5-6-9(10)14-11/h1-6H,(H2,12,13)
InChI key
FECQXVPRUCCUIL-UHFFFAOYSA-N
Biochem/physiol Actions
SKA-31 is an activator of KCa2 and KCa3.1 calcium-activated potassium channels.
SKA-31 is an activator of KCa2 and KCa3.1 calcium-activated potassium channels. Its EC50 is almost identical at 2-3 μM for all KCa2 subtypes, and about 10-fold lower (260 nM) at KCa3.1 channels. It has significant blood-pressure reducing activity in experimentally hypertensive mice, suggesting potassium channel activation as a therapeutic target for hypertension.
SKA31 is an activator of KCa2 and KCa3.1
SKA31 is an activator of KCa2 and KCa3.1 Potassium Channels, Potentiates the EDHF Response and Lowers Blood Pressure.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Alexander I Bondarenko et al.
Vascular pharmacology, 89, 39-48 (2017-01-09)
Lysophosphatidylinositol (LPI) and lysophosphatidylcholine (LPC) are lipid signaling molecules that induce endothelium-dependent vasodilation. In addition, LPC suppresses acetylcholine (Ach)-induced responses. We aimed to determine the influence of LPC and LPI on hyperpolarizing responses in vitro and in situ endothelial cells
Giuseppina D'Alessandro et al.
Cancers, 11(3) (2019-03-01)
Glioblastoma (GBM) is a deadly brain tumor, with fast recurrence even after surgical intervention, radio- and chemotherapies. One of the reasons for relapse is the early invasion of surrounding brain parenchyma by GBM, rendering tumor eradication difficult. Recent studies demonstrate
Sulie L Chang et al.
Journal of neuroimmune pharmacology : the official journal of the Society on NeuroImmune Pharmacology (2018-07-11)
Ethanol (EtOH), the main ingredient in alcoholic beverages, is well known for its behavioral, physiological, and immunosuppressive effects. There is evidence that EtOH acts through protein targets to exert its physiological effects; however, the mechanisms underlying EtOH's effects on inflammatory
Md A Hakim et al.
Journal of Alzheimer's disease : JAD, 76(4), 1423-1442 (2020-07-12)
Development of Alzheimer's disease (AD) pathology is associated with impaired blood flow delivery of oxygen and nutrients throughout the brain. Cerebrovascular endothelium regulates vasoreactivity of blood vessel networks for optimal cerebral blood flow. We tested the hypothesis that cerebrovascular endothelial
Lu Yun Zhang et al.
Frontiers in physiology, 12, 639857-639857 (2021-03-27)
Purposes: Since the role of store-operated calcium entry (SOCE) in endothelium-dependent hyperpolarization (EDH)-mediated vasorelaxation of mesenteric arteries in health and colitis is not fully understood, cyclopiazonic acid (CPA), a specific inhibitor of the sarco(endo) plasmic reticulum calcium-ATPases (SERCA), was used
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service