Skip to Content
Merck
All Photos(1)

Key Documents

S2438

Supelco

Streptavidin−Peroxidase Polymer, Ultrasensitive

Synonym(s):

Highly sensitive Streptavidin−Peroxidase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

form

buffered aqueous solution

Quality Level

shipped in

wet ice

storage temp.

−20°C

General description

Activated streptavidin (SA) and horseradish peroxidase (HRP) are covalently conjugated to a polymer backbone. Multiple active biomolecules on each polymer chain increase the biotin binding capacity and amplify the peroxidase enzyme signal.

Specificity

Biotinylated biomolecules.

Application

Detection of biotinylated proteins and nucleic acids and other biomolecules.
Suitable use in Western blotting and ELISA and for detecting surface antigens in immunohistochemistry and immunocytochemistry.

Physical form

Supplied as an 1.0 mg/ml solution in 0.01 M sodium phosphate, 0.15 M sodium chloride, 50% glycerol, stabilizer and preservative, pH 7.4.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Faruk Can et al.
Bioelectrochemistry (Amsterdam, Netherlands), 135, 107553-107553 (2020-05-23)
Applications of molecular techniques to elucidate identity or function using biomarkers still remain highly empirical and biosensors are no exception. In the present study, target-specific oligonucleotide probes for E. coli K12 were designed thermodynamically and applied in an electrochemical DNA
Natalia Gruba et al.
International journal of molecular sciences, 20(7) (2019-03-31)
Kallikrein 13 (KLK13) was first identified as an enzyme that is downregulated in a subset of breast tumors. This serine protease has since been implicated in a number of pathological processes including ovarian, lung and gastric cancers. Here we report
Aya Sugyo et al.
Cancers, 12(12) (2020-12-10)
In treatment-refractory cancers, tumor tissues damaged by therapy initiate the repair response; therefore, tumor tissues must be exposed to an additional burden before successful repair. We hypothesized that an agent recognizing a molecule that responds to anticancer treatment-induced tissue injury
Stefan Kaluz et al.
Biochemical and biophysical research communications, 370(4), 613-618 (2008-04-12)
The hypoxia-inducible factor (HIF) activates transcription via binding to the highly variable hypoxia-responsive elements (HREs). All hypoxia-inducible constructs described to date utilize multimers of naturally occurring HREs. Here, we describe the rational design of minimal hypoxia-inducible enhancers, conceptually equivalent to
Jaclyn C Law et al.
Journal of immunology (Baltimore, Md. : 1950), 206(1), 37-50 (2020-11-20)
There is a pressing need for an in-depth understanding of immunity to SARS-CoV-2. In this study, we investigated human T cell recall responses to fully glycosylated spike trimer, recombinant N protein, as well as to S, N, M, and E

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service