Skip to Content
Merck
All Photos(1)

Key Documents

S1445

Sigma-Aldrich

Anti-Somatostatin Receptor Type 1 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-SSTR1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 8 μg/mL using human pancreas, Islets of Langerhans

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... SSTR1(6751)
mouse ... Sstr1(20605)
rat ... Sstr1(25033)

General description

Somatostatin Receptor Type 1 (SSTR1) is present in blood vessels, neuroendocrine cells, and the retina. SSTR1 gene is mapped to human chromosome 14q21.1.

Immunogen

synthetic peptide corresponding to the C-terminal of human somatostatin receptor type 1, conjugated to KLH. The immunizing peptide has 100% homology with the rat and mouse gene.

Biochem/physiol Actions

Somatostatin Receptor Type 1 (SSTR1) protein extracellular loop regions are crucial for ligand binding. SSTR1 gene down-regulated in reported in preeclampsia but it is overexpressed in breast cancer. It is reported to block the mitogen-activated protein kinase (MAPK) pathway. However, in pancreatic cell lines, elevated expression of SSTR1 gene favors cell cycle arrest. SSTR1 also plays a tumor suppressor role in gastric cancer invasion.

Physical form

Solution in phosphate buffered saline, pH 7.7, containing 0.01% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Santhosh Kumar Nagarajan et al.
ACS omega, 5(33), 21145-21161 (2020-09-03)
Somatostatin receptor 1 (SSTR1), a subtype of somatostatin receptors, is involved in various signaling mechanisms in different parts of the human body. Like most of the G-protein-coupled receptors (GPCRs), the available information on the structural features of SSTR1 responsible for
J Zhao et al.
British journal of cancer, 108(12), 2557-2564 (2013-06-01)
Somatostatin receptor 1 (SSTR1) was preferentially methylated in Epstein-Barr virus (EBV)-positive gastric cancer using promoter methylation array. We aimed to analyse the epigenetic alteration and biological function of SSTR1 in EBV-associated gastric cancer (EBVaGC). Promoter methylation was examined by combined
Yi Zou et al.
Oncology letters, 17(2), 1723-1731 (2019-01-25)
Somatostatin receptors (SSTRs) are G-protein-coupled plasma membrane receptors that have been determined to be expressed in normal and cancer tissues. Activation of SSTRs frequently results in inhibition of cell proliferation and therefore somatostatin analogues (SSAs) have been used in cancer
Y-H Yan et al.
European review for medical and pharmacological sciences, 17(22), 3083-3094 (2013-12-05)
Preeclampsia is a complication of pregnancy that severely threatens the health of the mother and infant, yet the mechanism of pathogenesis remains unclear. In this article, gene array technology was applied to identify the genes related to the pathogenesis of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service