Skip to Content
Merck
All Photos(1)

Key Documents

S1389

Sigma-Aldrich

Anti-Sheep Red Blood Cell Stroma antibody produced in rabbit

fractionated antiserum, lyophilized powder

Synonym(s):

Anti-Sheep RBC Stroma, RBC Stroma Antibody

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

fractionated antiserum

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

sheep

packaging

vial of ~2 mL Lyophilized antiserum

technique(s)

agglutination assay: 1:100-1:200
immunoelectrophoresis: suitable
immunoelectrophoresis: suitable

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Immunogen

sheep red blood cell stroma (hemolysin)

Application

Anti-Sheep Red Blood Cell Stroma antibody produced in rabbit has been used in hemolysis assay.
Anti-Sheep Red Blood Cell Stroma antibody produced in rabbit was used in erythrocyte lysis assay.

Physical form

Lyophilized from 0.01 M phosphate buffered saline, pH 7.2

Reconstitution

Reconstitute with 2 ml deionized water.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Laure Yatime et al.
Frontiers in immunology, 9, 2822-2822 (2018-12-18)
The complement system is an efficient anti-microbial effector mechanism. On the other hand abnormal complement activation is involved in the pathogenesis of multiple inflammatory and hemolytic diseases. As general inhibition of the complement system may jeopardize patient health due to
Ilia A Krenev et al.
Marine drugs, 18(12) (2020-12-17)
Antimicrobial peptides (AMPs) are not only cytotoxic towards host pathogens or cancer cells but also are able to act as immunomodulators. It was shown that some human and non-human AMPs can interact with complement proteins and thereby modulate complement activity.
J Healer et al.
Infection and immunity, 67(5), 2334-2339 (1999-05-04)
Phagocytosis of Plasmodium falciparum sexual stages in vitro and within the mosquito midgut was assayed in order to assess its role in transmission-blocking immunity to malaria. Both monocytes/macrophages (MM) and polymorphonuclear neutrophils (PMN) phagocytosed malarial gametes in vitro, but levels
Annegret Wuensch et al.
Xenotransplantation, 27(5), e12585-e12585 (2020-02-15)
The activation of the endothelial surface in xenografts is still a poorly understood process and the consequences are unpredictable. The role of Ca2+ -messaging during the activation of endothelial cells is well recognized and routinely measured by synthetic Ca2+ -sensitive
Structural basis for therapeutic inhibition of complement C5
Jore MM, et al.
Nature Structural and Molecular Biology, 23(5), 378-378 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service