Skip to Content
Merck
All Photos(3)

Key Documents

S0532

Sigma-Aldrich

Strychnine

Synonym(s):

(−)-Strychnine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H22N2O2
CAS Number:
Molecular Weight:
334.41
Beilstein:
52979
EC Number:
MDL number:
UNSPSC Code:
12352210
PubChem Substance ID:
NACRES:
NA.54

Quality Level

mp

284-286 °C (lit.)

solubility

chloroform: soluble, clear to hazy

SMILES string

[H][C@@]12CC(=O)N3c4ccccc4[C@]56CC[N@H]7CC(=CCO1)[C@]([H])(C[C@@H]57)[C@]2([H])[C@]36[H]

InChI

1S/C21H22N2O2/c24-18-10-16-19-13-9-17-21(6-7-22(17)11-12(13)5-8-25-16)14-3-1-2-4-15(14)23(18)20(19)21/h1-5,13,16-17,19-20H,6-11H2/t13-,16-,17-,19-,20-,21+/m0/s1

InChI key

QMGVPVSNSZLJIA-FVWCLLPLSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Strychnine is a colorless, odourless and bitter tasting powder. It is absorbed in skin, gastrointestinaland respiratory tract. Strychnine generates oxides of nitrogen and carbon on decomposition.
Strychnine is a glycine receptor antagonist not associated with the N-methyl-D-aspartate (NMDA) receptor. It is an alkaloid extracted from Strychnos nux vomica seeds.

Application

Strychnine has been used as a glycine receptors antagonist in hippocampal CA1 pyramidal and rostral ventromedial medulla (RVM) neurons. It has also been used as type C glycine receptor antagonist in direction selective (DS) retinal cells.
Strychnine has been used in antagonist profiling to characterize a novel nicotinic receptor in Aplysia neuroendocrine cells.

Biochem/physiol Actions

At low doses, strychnine is used for circulatory problems, gastrointestinal disorders and eye diseases.
Strychnine is used as a rodenticide and a glycine receptor antagonist, which inhibits Renshaw cell-motor neuron synapse. It binds to the α subunit N-terminal region of the glycine receptor. The effect of strychnine is neutralized in the presence of glycine receptor agonist ivermectin. Strychnine also inhibits leukocyte motility.
Convulsant; glycine receptor antagonist not associated with the NMDA receptor.

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 1 Oral - Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Glycine-gated chloride channels in neutrophils attenuate calcium influx and superoxide production
WHEELER M, et al.
Faseb Journal, 14(3), 476-484 (2000)
Pharmacological Dissection of Intrinsic Optical Signal Reveals a Functional Coupling between Synaptic Activity and Astrocytic Volume Transient
Woo J, et al.
Experimental Neurobiology, 28(1), 30-42 (2019)
In vivo protection against strychnine toxicity in mice by the glycine receptor agonist ivermectin
Maher A, et al.
BioMed Research International (2014)
A brainstem-spinal cord inhibitory circuit for mechanical pain modulation by GABA and enkephalins
Franccois A, et al.
Neuron, 93(4), 822-839 (2017)
Spatiotemporally asymmetric excitation supports mammalian retinal motion sensitivity
Matsumoto A, et al.
Current Biology, 29(19) (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service