Skip to Content
Merck
All Photos(1)

Key Documents

S0193

Sigma-Aldrich

SKF-86002

≥98% (HPLC), solid

Synonym(s):

6-(4-Fluorophenyl)-5-(4-pyridyl)-2,3-dihydroimidazo[2,1-b]-thiazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H12N3FS
CAS Number:
Molecular Weight:
297.35
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

storage condition

protect from light

color

off-white

mp

189-190 °C (lit.)

solubility

DMSO: soluble >10 mg/mL

storage temp.

2-8°C

SMILES string

Fc1ccc(cc1)-c2nc3SCCn3c2-c4ccncc4

InChI

1S/C16H12FN3S/c17-13-3-1-11(2-4-13)14-15(12-5-7-18-8-6-12)20-9-10-21-16(20)19-14/h1-8H,9-10H2

InChI key

YOELZIQOLWZLQC-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

p38 MAP kinase inhibitor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Pouliot et al.
Journal of immunology (Baltimore, Md. : 1950), 158(10), 4930-4937 (1997-05-15)
Prostaglandin endoperoxide synthase (PGHS; cyclooxygenase), the rate-limiting enzyme in the conversion of arachidonic acid to prostanoids, has two isoforms. PGHS-1 is constitutively expressed and involved in homeostasis, whereas PGHS-2 is inducible in monocytes in response to proinflammatory agents. Using freshly
Maria Tsirigotis et al.
PloS one, 3(5), e2130-e2130 (2008-05-08)
The signaling pathways that may modulate the pathogenesis of diseases induced by expanded polyglutamine proteins are not well understood. Herein we demonstrate that expanded polyglutamine protein cytotoxicity is mediated primarily through activation of p38MAPK and that the atypical PKC iota
Donna E Muscarella et al.
Toxicological sciences : an official journal of the Society of Toxicology, 68(1), 82-92 (2002-06-21)
Various forms of inorganic arsenic are significant environmental contaminants that have multiple effects on cells, including the induction of apoptotic cell death. Induction of apoptosis in lymphoid cells can mediate immunotoxicity following exposure to chemicals. However, the mechanisms regulating the
Gerd Wagner et al.
Medicinal research reviews, 26(1), 1-62 (2005-11-12)
The recent successful introduction of the anti-cytokine biologicals Etanercept, Infliximab, Adalimumab, and Anakinra has stimulated the search for anti-cytokine small-molecules. A number of molecular targets have been identified for the development of such small molecular anti-cytokine agents. The focus of
T Ishizuka et al.
Proceedings of the National Academy of Sciences of the United States of America, 94(12), 6358-6363 (1997-06-10)
Mast cells synthesize and secrete specific cytokines and chemokines which play an important role in allergic inflammation. Aggregation of the high-affinity Fc receptor (FcepsilonRI) for immunoglobulin E (IgE) in MC/9 mouse mast cells stimulates the synthesis and secretion of tumor

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service