Skip to Content
Merck
All Photos(1)

Key Documents

RAB1092

Sigma-Aldrich

Human TSG-6 ELISA Kit

for cell culture supernatants, plasma, and serum samples

Synonym(s):

Hyaluronate-binding protein, TSG-6, Tumor necrosis factor-inducible gene 6 protein 

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type cell culture supernatant(s)
sample type serum sample(s)
sample type plasma

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.2 ng/ml

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... TNFAIP6(7130)

General description

This ELISA antibody pair detects human TSG-6. Other species not yet determined.

Application

Human TSG-6 ELISA Kit has been used to determine galectin G3 and intermediate metabolites level.

Biochem/physiol Actions

Tumor necrosis factor (TNF)-α–stimulated gene 6 protein (TSG-6) has been implicated in regulation of cell-cell and cell-matrix interactions during inflammation and tumorigenesis. TSG-6 possess anti-inflammatory properties. It is also known to reduce inflammatory reactions triggered by ischemia in the heart. Thus, it might have a potential role in treating heart disease with other cells such as skeletal myoblasts, fetal myoblasts and embryonic stem cells.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Keisin Wang et al.
The Journal of surgical research, 220, 311-319 (2017-11-29)
The formation of abdominal aortic aneurysms (AAA) is characterized by a dominance of proinflammatory forces that result in smooth muscle cell apoptosis, extracellular matrix degradation, and progressive diameter expansion. Additional defects in the antiinflammatory response may also play a role
Hao Yang et al.
Journal of neuroinflammation, 17(1), 154-154 (2020-05-13)
Neuroinflammation plays a vital role in the development and maintenance of neuropathic pain. Recent evidence has proved that bone marrow mesenchymal stem cells (BMSCs) can inhibit neuropathic pain and possess potent immunomodulatory and immunosuppressive properties via secreting a variety of
Anti-inflammatory protein TSG-6 secreted by activated MSCs attenuates zymosan-induced mouse peritonitis by decreasing TLR2/NF-?B signaling in resident .
Hosoo , et al.
Blood, 118, 330-338 (2011)
Chao Yang et al.
Molecular and cellular biochemistry, 446(1-2), 171-184 (2018-01-23)
Mesenchymal stem cells (MSCs) are used as therapeutic tool for the treatment of immune diseases. The inflammatory environment also influences the characteristics of MSCs after transplantation. The aim of the study was to investigate the effects of pro-inflammatory cytokines on
Milad Riazifar et al.
ACS nano, 13(6), 6670-6688 (2019-05-24)
To dissect therapeutic mechanisms of transplanted stem cells and develop exosome-based nanotherapeutics in treating autoimmune and neurodegenerative diseases, we assessed the effect of exosomes secreted from human mesenchymal stem cells (MSCs) in treating multiple sclerosis using an experimental autoimmune encephalomyelitis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service