Skip to Content
Merck
All Photos(1)

Key Documents

RAB0804

Sigma-Aldrich

Mouse IgG2b ELISA Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.84

species reactivity

mouse

technique(s)

ELISA: suitable

assay range

inter-assay cv: <10%
intra-assay cv: <12%

shipped in

wet ice

storage temp.

−20°C

General description

The antibody pair provided in this kit recognizes Mouse IgG2b.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Niloufar Kavian et al.
Clinical & translational immunology, 9(2), e1107-e1107 (2020-02-07)
Enhanced inactivated influenza vaccines (eIIV) aim to increase immunogenicity and protection compared with the widely used standard IIV (S-IIV). We tested four vaccines in parallel, FluZone high dose, FluBlok and FluAd versus S-IIV in a randomised controlled trial of older
Zhexin Fan et al.
Food & function, 11(5), 4499-4511 (2020-05-10)
Emerging studies have addressed the role of probiotics in inflammation modulation via modifying gut microbiota. Perturbed gut microbiota is recognized as a pivotal trigger in the pathogenesis of rheumatoid arthritis (RA), and manipulating gut microbiota at the early phase may
Cecilia A Langellotti et al.
Viral immunology, 34(2), 68-78 (2020-11-05)
Bovine herpesvirus-1 (BoHV-1) uses many mechanisms to elude the immune system; one of them is spreading intracellularly, even in the presence of specific antiviral antibodies. Cytotoxic T lymphocytes (CTLs) are necessary to eliminate the virus. The main preventive strategy is
Xiangguo Duan et al.
Frontiers in immunology, 11, 701-701 (2020-05-16)
Follicular helper T (Tfh) cells regulate high-affinity antibody production. Some findings have indicated that Tfh cells could be differentiated into memory cells. Here we have investigated the effects of IFN-α, as an adjuvant, on the generation of memory Tfh cell
Xingxing Jian et al.
Microbiome, 8(1), 74-74 (2020-05-30)
Gut microbiome alterations are closely related to human health and linked to a variety of diseases. Although great efforts have been made to understand the risk factors for multiple myeloma (MM), little is known about the role of the gut

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service