Skip to Content
Merck
All Photos(1)

Key Documents

RAB0751

Sigma-Aldrich

Human UMOD / Uromodulin ELISA Kit

Synonym(s):

THP, Tamm-Horsfall protein, Tamm-Horsfall urinary glycoprotein, Uromodulin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.84

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type serum
sample type plasma
sample type cell culture supernatant(s)

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 40 pg/mL
standard curve range: 40.96-10000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... UMOD(7369)

General description

The UMOD ELISA kit provides for the Quantitative measurement of Uromodulin in Cell Culture Supernatants, Plasma and Serum.
Uromodulin also known as Tamm-Horsfall protein (THP) is a glycoprotein localized in the thick ascending limb (TAL) of the Henle loop in the kidney. It is the most abundant protein excreted in the urine. Uromodulin plays a role in preventing urinary tract infections and kidney stones. It aids in immunomodulation and regulation of ion transport in TAL. Uromodulin acts as an inverse biomarker in the early detection of chronic kidney disorders.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Human UMOD / Uromodulin ELISA Kit has been used to measure the levels of Tamm-Horsfall protein (THP) in human plasma samples.

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Olivier Devuyst et al.
Nature reviews. Nephrology, 13(9), 525-544 (2017-08-07)
Uromodulin (also known as Tamm-Horsfall protein) is exclusively produced in the kidney and is the most abundant protein in normal urine. The function of uromodulin remains elusive, but the available data suggest that this protein might regulate salt transport, protect
Kaice A LaFavers et al.
Science translational medicine, 11(512) (2019-10-04)
High serum concentrations of kidney-derived protein uromodulin [Tamm-Horsfall protein (THP)] have recently been shown to be independently associated with low mortality in both older adults and cardiac patients, but the underlying mechanism remains unclear. Here, we show that THP inhibits

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service