Skip to Content
Merck
All Photos(1)

Key Documents

RAB0521

Sigma-Aldrich

Bovine IL2 / Interleukin-2 ELISA Kit

for serum, plasma and cell culture supernatants

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32
Pricing and availability is not currently available.

species reactivity

bovine

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type serum
sample type cell culture supernatant(s)
sample type plasma

assay range

inter-assay cv: <10%
intra-assay cv: <12%
sensitivity: 0.2 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

bovine ... IL2(280822)

General description

This ELISA antibody pair detects bovine Interleukin-2.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)

Other Notes

A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.

Kit Components Also Available Separately

Product No.
Description
SDS

  • RABTMB3ELISA Colorimetric TMB Reagent (HRP Substrate, Item H)SDS

  • RABSTOP3ELISA Stop Solution (Item I)SDS

  • RABELADBELISA 5X Assay/Sample Diluent Buffer B (Item E1)SDS

  • RABELADCELISA 1X Assay/Sample Diluent Buffer C (Item L)SDS

  • RABWASH420X Wash Buffer (Item B)SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liping Qiu et al.
Small (Weinheim an der Bergstrasse, Germany), 14(15), e1703539-e1703539 (2018-03-02)
The activation of tumor-specific effector immune cells is key for successful immunotherapy and vaccination is a powerful strategy to induce such adaptive immune responses. However, the generation of effective anticancer vaccines is challenging. To overcome these challenges, a novel straight-forward
Francesco Nannini et al.
Scientific reports, 10(1), 19168-19168 (2020-11-07)
Antibody phage display is a powerful platform for discovery of clinically applicable high affinity monoclonal antibodies against a broad range of targets. Libraries generated from immunized animals offer the advantage of in vivo affinity-maturation of V regions prior to library
Pratiksha Gulati et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 24(16), 3981-3993 (2018-05-12)
Purpose: Combination therapy of adoptively transferred redirected T cells and checkpoint inhibitors aims for higher response rates in tumors poorly responsive to immunotherapy like malignant pleural mesothelioma (MPM). Only most recently the issue of an optimally active chimeric antigen receptor
Tatsuo Matsuda et al.
Clinical cancer research : an official journal of the American Association for Cancer Research, 24(21), 5357-5367 (2018-05-04)
Purpose: Current evolution of cancer immunotherapies, such as immune checkpoint blockade, has implicated neoantigens as major targets of anticancer cytotoxic T cells. Adoptive T-cell therapy with neoantigen-specific T-cell receptor (TCR)-engineered T cells would be an attractive therapeutic option for advanced
Zhengping Hu et al.
Scientific reports, 8(1), 217-217 (2018-01-11)
The increased PD-L1 induces poorer prognosis in melanoma. The treatment with PD-1/PD-L1 antibodies have a low response rate. The combination immunotherapies are the encouraging drug development strategy to receive maximal therapeutic benefit. In this study, we investigated the enhanced antitumor

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service