RAB0431
Mouse Shh-N ELISA Kit
for serum, plasma and cell culture supernatant
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
mouse
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type cell culture supernatant(s)
sample type serum
sample type plasma
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 5 pg/mL
standard curve range: 4.1-1000 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
mouse ... Shh(20423)
General description
The Mouse Shh-N (Sonic Hedgehog N Terminus) ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse Shh-N in serum, plasma, cell culture supernatants and urine.
Immunogen
Recombinant Mouse Sonic Hedgehog (N-terminus)
Application
For research use only. Not for use in diagnostic procedures.
Mouse Shh-N ELISA Kit has been used to measure serum sonic hedgehog (Shh) level.
Mouse Shh-N ELISA Kit has been used to measure serum sonic hedgehog (Shh) level.
Biochem/physiol Actions
Sonic hedgehog (Shh) is a secreted glycoprotein involved in embryonic morphogenic development of several tissues. It has a role in fracture healing and regeneration. In the canonical pathway, the protein associates with its receptor Patched (Ptc-1), de-represses Smoothened (Smo) and activates the Gli family of transcription factors. Mutations in the Shh pathway have been linked to developmental diseases and cancer. Shh plays a vital role in axon growth and guidance throughout the brain development. Shh stimulates neurite outgrowth by overcoming neurite growth inhibitory environment associated with glial scar.
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Reduced lipoapoptosis, hedgehog pathway activation and fibrosis in caspase-2 deficient mice with non-alcoholic steatohepatitis.
Machado MV
Gut, 64(7), 1148-1157 (2015)
An epigenetic switch induced by Shh signalling regulates gene activation during development and medulloblastoma growth.
Shi X
Nature Communications, 5:5425-5:5425 (2014)
Sonic hedgehog stimulates neurite outgrowth in a mechanical stretch model of reactive-astrogliosis.
Berretta A
Scientific Reports, 6:21896-6:21896 (2016)
M V Machado et al.
Gut, 64(7), 1148-1157 (2014-07-24)
Caspase-2 is an initiator caspase involved in multiple apoptotic pathways, particularly in response to specific intracellular stressors (eg, DNA damage, ER stress). We recently reported that caspase-2 was pivotal for the induction of cell death triggered by excessive intracellular accumulation
Lin Ding et al.
Cellular and molecular gastroenterology and hepatology, 11(5), 1251-1266 (2020-12-22)
Helicobacter pylori infection in humans typically begins with colonization of the gastric antrum. The initial Th1 response occasionally coincides with an increase in gastrin secretion. Subsequently, the gastritis segues to chronic atrophic gastritis, metaplasia, dysplasia and distal gastric cancer. Despite
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service