Skip to Content
Merck
All Photos(1)

Key Documents

RAB0385

Sigma-Aldrich

Atrial Natriuretic Peptide EIA Kit

for serum, plasma, culture supernatant and cell lysates

Synonym(s):

Natriuretic peptides A, Prepronatriodilatin, ANP

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.32

species reactivity

mouse, human, rat

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable
enzyme immunoassay: suitable

input

sample type serum
sample type cell lysate
sample type plasma
sample type culture supernatant(s)

assay range

inter-assay cv: <15%
intra-assay cv: <10%
sensitivity: 1.02 pg/mL
standard curve range: 0.1-1000 pg/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... NPPA(4878)

General description

The ANP Enzyme Immunoassay (EIA) Kit is an in vitro quantitative assay for detecting ANP peptide based on the principle of Competitive Enzyme Immunoassay. EIA-ANP-1 targets the common sequence of human, mouse and rat, and thus may be used to detect ANP expression in all these species with high specificity and sensitivity.

Immunogen

Atrial Natriuretic Peptide, synthetic peptide

Application

Atrial Natriuretic Peptide EIA Kit has been used for the quantification of atrial natriuretic peptide (ANP) in cell culture.
For research use only. Not for use in diagnostic procedures.
Atrial Natriuretic Peptide EIA Kit has been used to measure ANP levels in diabetes mellitus patients.

Biochem/physiol Actions

Atrial natriuretic peptide (ANP) is a polypeptide hormone released in response to atrial dilation and it stimulates diuresis, natriuresis and vasorelaxation. ANP acts as an inhibitor of renin-angiotensin-aldosterone system (RAAS) pathway. It also has a major role in body fluid homeostasis and maintenance of blood pressure. Binding of ANP to receptors on target cell stimulate the production of cyclic guanosine monophosphate (GMP). Overexpression of ANP leads to congestive heart failure in the human. Experimental studies show that ANP can be used as a therapeutic for stimulating angiogenesis in ischemic tissues. N-terminal ANP prohormone functions as a biomarker for asymptomatic systolic left ventricular (LV) dysfunction, congestive heart failure and cardiovascular mortality.

Kit Components Also Available Separately

Product No.
Description
SDS

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Natriuretic peptides: Diagnostic and therapeutic use.
Pandit K
Indian Journal of Endocrinology and Metabolism, suppl 4, S345-S353 (2011)
Joseph A Hippensteel et al.
Critical care (London, England), 23(1), 259-259 (2019-07-25)
Intravenous fluids, an essential component of sepsis resuscitation, may paradoxically worsen outcomes by exacerbating endothelial injury. Preclinical models suggest that fluid resuscitation degrades the endothelial glycocalyx, a heparan sulfate-enriched structure necessary for vascular homeostasis. We hypothesized that endothelial glycocalyx degradation
Atrial natriuretic peptide elevation in congestive heart failure in the human.
Burnett JC Jr
Science, 231(4742), 1145-1147 (1986)
Endothelial cell modulation of cardiomyocyte gene expression
Jiang F, et al.
Experimental Cell Research, 383(2), 111565-111565 (2019)
Atrial natriuretic peptide in heart failure.
Brandt RR
Journal of the American College of Cardiology, 22(4 suppl A), 86A-92A (1993)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service