RAB0231
Mouse IGF-II ELISA Kit
for cell culture supernatant
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
mouse
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type cell culture supernatant(s)
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 1500 pg/mL
standard curve range: 2.3-150 ng/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
mouse ... Igf2(16002)
General description
The Mouse IGF-II ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of mouse IGF-II in serum, plasma, cell culture supernatants and urine.
Immunogen
Recombinant Mouse IGF-II
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8B - Non-combustible corrosive hazardous materials
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Higher Maternal Protein Intake during Pregnancy Is Associated with Lower Cord Blood Concentrations of Insulin-like Growth Factor (IGF)-II, IGF Binding Protein 3, and Insulin, but Not IGF-I, in a Cohort of Women with High Protein Intake.
Karen M Switkowski et al.
The Journal of nutrition, 147(7), 1392-1400 (2017-06-09)
Davide Serrano et al.
Nutrients, 13(2) (2021-01-29)
Obesity and diet are associated with colorectal cancer (CRC) risk, and microbiome could mediate this risk factor. To investigate this interaction, we performed a case-control study (34 CRC cases and 32 controls) and analyzed fecal microbiota composition using 16S rRNA
Mototsugu Nagao et al.
European journal of endocrinology, 184(1), 41-49 (2020-10-29)
In insulin-like growth factor II (IGF-II) producing non-islet cell tumor hypoglycemia (NICTH), high molecular weight forms of IGF-II (big IGF-II) are produced as a cause of spontaneous hypoglycemia. MicroRNA (miRNA)-483 family, encoded in an intron lesion of IGF2 gene, is
Jun Yang et al.
BMC medicine, 18(1), 268-268 (2020-10-07)
Pulmonary arterial hypertension (PAH) is a fatal disease that results from cardio-pulmonary dysfunction with the pathology largely unknown. Insulin-like growth factor binding protein 2 (IGFBP2) is an important member of the insulin-like growth factor family, with evidence suggesting elevation in
Meng-Nan Yang et al.
Pediatric research, 87(5), 946-951 (2019-12-01)
Retinol-binding protein 4 (RBP-4) is an adipokine involved in regulating insulin sensitivity which would affect fetal growth. It is unclear whether RBP-4 is associated with fetal overgrowth, and unexplored which fetal growth factor(s) may mediate the association. In the Shanghai
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service