RAB0098
Human M-CSF / CSF1 ELISA Kit
for serum, plasma, cell culture supernatant, urine
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
human
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type plasma
sample type serum
sample type cell culture supernatant(s)
sample type urine
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 5 pg/mL
standard curve range: 2.78-2000 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
human ... CSF1(1435)
General description
M-CSF (colony stimulating factor 1) is a pleiotropic macrophage growth factor. It has a signal peptide, a 149-amino acid receptor-binding domain, a variable spacer region and a transmembrane region with a cytoplasmic tail. The human M-CSF receptor is a 972-amino acid glycosylated protein which consist of an extracellular region made of five immunoglobulin-like domains (D1–D5), a transmembrane region and an intracellular autocatalytic kinase domain. This gene is mapped to human chromosome p13-p21.
The Human MCSF ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of human MCSF in serum, plasma, cell culture supernatants and urine.
Immunogen
Recombinant Human MCSF
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Biochem/physiol Actions
M-CSF (colony stimulating factor 1) plays a major role in the immune response, bone metabolism and the development of some cancers. It modulates macrophage survival, proliferation, differentiation and chemotaxis.
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Combinatorial and computational approaches to identify interactions of macrophage colony-stimulating factor (M-CSF) and its receptor c-FMS.
Rosenfeld L, et al.
The Journal of Biological Chemistry, 290(43), 26180-26193 (2015)
Reassignment of the human CSF1 gene to chromosome 1p13-p21.
Morris SW, et al.
Blood, 78(8), 2013-2020 (1991)
Teresa Di Desidero et al.
Pharmacological research, 158, 104920-104920 (2020-05-29)
Anaplastic thyroid cancer (ATC) is a rare neoplasia with a poor prognosis. Proliferation and apoptosis assays were performed on ATC cell lines (8305C, 8505C) exposed to vinorelbine, lenvatinib, as well as to concomitant combinations. ABCB1, ABCG2 and CSF-1 mRNA expression
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service