RAB0057
Rat MCP-1 / CCL2 ELISA Kit
for serum, plasma and cell culture supernatant
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41116158
NACRES:
NA.32
Recommended Products
species reactivity
rat
packaging
kit of 96 wells (12 strips x 8 wells)
technique(s)
ELISA: suitable
capture ELISA: suitable
input
sample type plasma
sample type cell culture supernatant(s)
sample type serum
assay range
inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 15 pg/mL
standard curve range: 24.67-18000 pg/mL
detection method
colorimetric
shipped in
wet ice
storage temp.
−20°C
Gene Information
rat ... Ccl2(24770)
General description
The Rat MCP-1 ELISA (Enzyme-Linked Immunosorbent Assay) kit is an in vitro enzyme-linked immunosorbent assay for the quantitative measurement of rat MCP-1 in serum, plasma, cell culture supernatants and urine.
Immunogen
Recombinant Rat MCP1
Application
For research use only. Not for use in diagnostic procedures.
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Please refer to the attached General ELISA KIT Procedure (sandwich, competitive & Indirect ELISA)
Other Notes
A sample Certificate of Analysis is available for this product.
Please type the word sample in the text box provided for lot number.
Please type the word sample in the text box provided for lot number.
Kit Components Also Available Separately
Product No.
Description
SDS
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Met. Corr. 1
Storage Class Code
8A - Combustible corrosive hazardous materials
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Ammar J Alsheikh et al.
American journal of physiology. Renal physiology, 318(4), F982-F993 (2020-03-10)
Studies examining mechanisms of Dahl salt-sensitive (SS) hypertension have implicated the infiltration of leukocytes in the kidneys, which contribute to renal disease and elevated blood pressure. However, the signaling pathways by which leukocytes traffic to the kidneys remain poorly understood.
Ruozhi Zhao et al.
The Journal of nutritional biochemistry, 25(8), 851-857 (2014-06-14)
Atherosclerotic cardiovascular complications are the leading cause of death in diabetic patients. Monocyte adhesion is an early event for atherogenesis. Previous studies demonstrated that dark-skin berries had cardiovascular protective effects. We hypothesize that Saskatoon berry (SB) powder may reduce monocyte
Zhenyu Yao et al.
Journal of biomedical materials research. Part A, 102(9), 3291-3297 (2013-10-15)
Wear particles generated from total joint replacements can stimulate macrophages to release chemokines, such as monocyte chemoattractant protein 1 (MCP-1), which is the most important chemokine regulating systemic and local cell trafficking and infiltration of monocyte/macrophages in chronic inflammation. One
Sabine Kahl et al.
Diabetologia, 57(10), 2094-2102 (2014-07-23)
The combined IVGTT-hyperinsulinaemic-euglycaemic clamp (Botnia clamp) allows the assessment of insulin secretion and sensitivity in one experiment. It remains unclear whether this clamp yields results comparable with those of the standard hyperinsulinaemic-euglycaemic clamp (SHEC) in diabetes patients. We hypothesised that
John E S Park et al.
Thorax, 69(12), 1120-1127 (2014-09-17)
The burden of chronic heart failure (HF) is rising owing to an increased survivorship after myocardial infarction (MI). Pulmonary structural remodelling in patients with HF may protect against oedema while causing dyspnoea, the predominant symptom associated with HF. The cellular
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service