Skip to Content
Merck
All Photos(1)

Key Documents

R9001

Sigma-Aldrich

Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae)

Type X, lyophilized powder

Synonym(s):

Transfer RNA, tRNA

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
41106305
NACRES:
NA.55

grade

for molecular biology

Quality Level

type

Type X

form

lyophilized powder

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Transfer RNA (tRNA) is isolated from baker′s yeast by phenol-chloroform extraction and ethanol precipitation.

Application

Suitable for use as a carrier in nucleic acid purification and precipitation.

Components

tRNA is provided as a lyophilized powder.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Christophe Maris et al.
Nucleic acids research, 48(8), 4521-4537 (2020-03-15)
The polypyrimidine tract binding protein (PTB) is a multi-domain protein involved in alternative splicing, mRNA localization, stabilization, polyadenylation and translation initiation from internal ribosome entry sites (IRES). In this latter process, PTB promotes viral translation by interacting extensively with complex
A simplified method for combined immunohistochemistry and in-situ hybridization in fresh-frozen, cryocut mouse brain sections.
Newton, S.S., et al.
Brain Res. Protoc., 9, 214-214 (2002)
Mammalian in vitro splicing assays.
A Mayeda et al.
Methods in molecular biology (Clifton, N.J.), 118, 315-321 (1999-11-05)
Rahul Sharma et al.
STAR protocols, 1(1), 100006-100006 (2020-10-29)
DNA-FISH remains the method of choice to visualize genomic regions in situ ranging from a single locus to entire chromosomes. Current methods to generate probes rely on expensive kits that vary in labeling efficiency and are limited by the size
Andreas M Anger et al.
Nature, 497(7447), 80-85 (2013-05-03)
Protein synthesis in all cells is carried out by macromolecular machines called ribosomes. Although the structures of prokaryotic, yeast and protist ribosomes have been determined, the more complex molecular architecture of metazoan 80S ribosomes has so far remained elusive. Here

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service