Skip to Content
Merck
All Photos(1)

Key Documents

R8154

Sigma-Aldrich

Monoclonal Anti-RAP1 antibody produced in mouse

~2 mg/mL, clone 4C8/1, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-TERF2-Interacting Protein, Anti-TERF2IP

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4C8/1, monoclonal

form

buffered aqueous solution

mol wt

antigen ~47 kDa

species reactivity

human

concentration

~2 mg/mL

technique(s)

indirect ELISA: suitable
microarray: suitable
western blot: 0.1-0.2 μg/mL using HeLa total cell extract.

isotype

IgG2b

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TERF2IP(54386)

General description

Monoclonal Anti-RAP1 (mouse IgG2b isotype) is derived from the hybridoma 4C8/1 produced by the fusion of mouse myeloma cells (Sp2/0 Ag.14 cells) and splenocytes from BALB/c mice immunized with human RAP1. Ras-related protein 1 (RAP1) protein contains 399 amino acids with an acidic C-terminus featuring a 33 amino acid coiled-coil region and a bipartite nuclear localization signal. It is expressed ubiquitously, is located at telomeres and affects telomer length.
Rap1 (repressor/activator protein) is a constituent of shelterin. It consists of an N-terminal BRCT motif, a C-terminal protein interaction domain, and one or two central Myb domains. It is also known as Terf2ip (TERF2 interacting protein). It is located on chromosome 16q23.1.

Immunogen

human RAP1.

Application

Monoclonal Anti-RAP1 antibody produced in mouse may be used in enzyme linked immunosorbent assay (ELISA) and immunoblotting.

Biochem/physiol Actions

Rap1 (repressor/activator protein) helps to guard telomeres, assemble telomerase, and controls telomere length. At telomeres, Rap1 also participates in the suppression of non-homologous end joining (NHEJ). It involves in genomewide transcriptional modulation. Knockdown of Rap1 induces apoptosis in hepatocellular carcinoma cell line HepG2.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Genetic variants in eleven telomere-associated genes and the risk of incident cardio/cerebrovascular disease: The Women's Genome Health Study
Zee RY, et al.
Clinica Chimica Acta; International Journal of Clinical Chemistry, 412(1-2), 199-202 (2011)
TALEN gene knockouts reveal no requirement for the conserved human shelterin protein Rap1 in telomere protection and length regulation
Kabir S, et al.
Cell Reports, 9(4), 1273-1280 (2014)
Downregulation of Rap1 promotes 5-fluorouracil-induced apoptosis in hepatocellular carcinoma cell line HepG2
Zha Y, et al.
Oncology Reports, 31(4), 1691-1698 (2014)
Decoding telomere protein Rap1: Its telomeric and nontelomeric functions and potential implications in diabetic cardiomyopathy
Cai Y, et al.
Cell Cycle, 16(19), 1765-1765 (2017)
Mammalian RAP1 controls telomere function and gene expression through binding to telomeric and extra-telomeric sites
Martinez P, et al.
Nature Cell Biology, 12(8), 768?780-768?780 (2010)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service