Skip to Content
Merck
All Photos(5)

Key Documents

R7904

Sigma-Aldrich

Anti-Rab5 antibody, Mouse monoclonal

clone Rab5-65, purified from hybridoma cell culture

Synonym(s):

Anti-RAS-associated Protein RAB5A

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

Rab5-65, monoclonal

form

buffered aqueous solution

mol wt

~24 kDa

species reactivity

human, cat, bovine, mouse, rat

concentration

~2 mg/mL

technique(s)

immunocytochemistry: 5-10 μg/mL using HeLa cells or NIH-3T3 or NRK
indirect ELISA: suitable
microarray: suitable
western blot: suitable

isotype

IgG3

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RAB5A(5868)
mouse ... Rab5a(271457)
rat ... Rab5a(64633)

General description

Anti-Rab5 antibody, Mouse monoclonal (mouse IgG3 isotype) is derived from the hybridoma Rab5-65 produced by the fusion of mouse myeloma cells (NS1 cells) and splenocytes from BALB/c mice immunized with a synthetic peptide corresponding to C-terminal region of human Rab5, conjugated to KLH. Rab5 is ubiquitously expressed in human tissues. It localizes mainly to early endosomes, but is also present on the plasma membrane.
Rab5 (Ras-related protein Rab-5A) is a small GTPase , that has three isoforms - Rab5A, Rab5B and Rab5C. It is upregulated in several secretory cells. It is located on human chromosome 3p24.

Immunogen

synthetic peptide corresponding to the C- terminal region of human Rab5, conjugated to KLH.

Application

Anti-Rab5 antibody, Mouse monoclonal has been used in:
  • enzyme linked immunosorbent assay (ELISA)
  • immunocytochemistry
  • co-immunoprecipitation assay
  • western blotting
  • immunofluorescence

Biochem/physiol Actions

Among the proteins recruited by the GTP-bound active Rab5 are Rabaptin-5 and early endosome antigen 1(EEA1).
Rab5 (Ras-related protein Rab-5A) controls endosomal trafficking. Overexpression of Rab5A leads to metastasis and poor prediction in colorectal cancer patients. It modulates common Rab5 effectors to control the secretory pathways. It controls the homotypic integration between early endosomes and also the integration between endocytic vesicles and early endosomes.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pamela A Godoy et al.
Biomedicine & pharmacotherapy = Biomedecine & pharmacotherapie, 142, 111968-111968 (2021-08-04)
Amyloid beta peptide (Aβ) is tightly associated with the physiopathology of Alzheimer's Disease (AD) as one of the most important factors in the evolution of the pathology. In this context, we previously reported that Aβ increases the expression of ionotropic
Probing the druggability of membrane-bound Rab5 by molecular dynamics simulations
Edler E and Stein M
Journal of Enzyme Inhibition and Medicinal Chemistry, 32(1), 434-443 (2017)
The Autophagic Process Occurs in Human Bone Metastasis and Implicates Molecular Mechanisms Differently Affected by Rab5a in the Early and Late Stages
Maroni P, et al.
International Journal of Molecular Sciences, 17(4) (2016)
Thrombospondin expression in myofibers stabilizes muscle membranes
Vanhoutte D, et al.
eLife, 5, e17589-e17589 (2016)
Peng Zhao et al.
Biochimica et biophysica acta. General subjects, 1863(2), 502-510 (2018-12-12)
Magnetic nanoparticles (MNPs) have been widely used in biomedical applications. Proper control of the duration of MNPs in circulation promises to improve further their applications, in particularly drug delivery. It is known that the uptake of tissue-associated MNPs is mainly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service