Skip to Content
Merck
All Photos(1)

Key Documents

R7256

Sigma-Aldrich

RPMI 1640 Vitamins Solution (100×)

liquid, sterile-filtered, BioReagent, suitable for cell culture

Synonym(s):

RPMI 1640 Solution, RPMI 1640 Vitamins, Vitamins Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352205
NACRES:
NA.75

sterility

sterile-filtered

Quality Level

product line

BioReagent

form

liquid

technique(s)

cell culture | mammalian: suitable

impurities

endotoxin, tested

color

yellow to dark yellow

pH

7.2-7.6

shipped in

dry ice

storage temp.

−20°C

General description

Roswell Park Memorial Institute medium is commonly referred as RPMI. It is a form of medium used in cell culture and tissue culture. This medium contains phosphate and is formulated for use in a 5% carbon dioxide atmosphere. RPMI 1640 is used for the serum-free expansion of human lymphoid cells.

Application

RPMI 1640 Vitamins Solution (100×) has been used:
  • in the preparation of CeM1 medium for the isolation of Caenorhabditis elegans germ cells
  • as a component of the amino-acid-deprived medium for the starvation of intestinal porcine enterocytes
  • as a component of the stage-8 medium for culturing of human pluripotent stem cell-derived 3D pancreatic progenitor clusters
RPMI 1640 Vitamins Solution (100×) is used as a supplement in cell culture media.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leucine stimulates ASCT2 amino acid transporter expression in porcine jejunal epithelial cell line (IPEC-J2) through PI3K/Akt/mTOR and ERK signaling pathways
Shihai Z, et al.
Amino Acids, 46(12) (2014)
Indranil Biswas et al.
Journal of bacteriology, 189(18), 6521-6531 (2007-07-10)
Streptococcus mutans, the principal causative agent of dental caries, produces four glucan-binding proteins (Gbp) that play major roles in bacterial adherence and pathogenesis. One of these proteins, GbpC, is an important cell surface protein involved in biofilm formation. GbpC is
Joshua R Kane et al.
eLife, 9 (2020-01-22)
In pursuit of therapeutics for human polyomaviruses, we identified a peptide derived from the BK polyomavirus (BKV) minor structural proteins VP2/3 that is a potent inhibitor of BKV infection with no observable cellular toxicity. The thirteen-residue peptide binds to major
Alexandra S Vagasi et al.
Bio-protocol, 7(15) (2017-09-05)
The Caenorhabditis elegans germ line is an important model system for the study of germ stem cells. Wild-type C. elegans germ cells are syncytial and therefore cannot be isolated in in vitro cultures. In contrast, the germ cells from tumorous
Nieves Vizcaíno et al.
Veterinary research, 51(1), 101-101 (2020-08-17)
Brucella ovis is a non-zoonotic rough Brucella that causes genital lesions, abortions and increased perinatal mortality in sheep and is responsible for important economic losses worldwide. Research on virulence factors of B. ovis is necessary for deciphering the mechanisms that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service