Skip to Content
Merck
All Photos(1)

Key Documents

R4758

Sigma-Aldrich

Resorufin β-D-glucopyranoside

≥90%, powder

Synonym(s):

3-Phenoxazone 7-(β-D-glucopyranoside)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H17NO8
CAS Number:
Molecular Weight:
375.33
Beilstein:
4335306
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

product name

Resorufin β-D-glucopyranoside, ≥90%

Quality Level

Assay

≥90%

form

powder

solubility

DMSO: 20 mg/mL

storage temp.

−20°C

SMILES string

OC[C@H]1O[C@@H](Oc2ccc3N=C4C=CC(=O)C=C4Oc3c2)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C18H17NO8/c20-7-14-15(22)16(23)17(24)18(27-14)25-9-2-4-11-13(6-9)26-12-5-8(21)1-3-10(12)19-11/h1-6,14-18,20,22-24H,7H2/t14-,15-,16+,17-,18-/m1/s1

InChI key

QULZFZMEBOATFS-UYTYNIKBSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Resorufin β-D-glucopyranoside has been used:
  • as a substrate to determine the kinetics properties for Clostridium cellulovorans (CcBglA), the fungus Trichoderma reesei (TrBgl2), and the termite Neotermes koshunensis (NkBg)
  • as a chromogenic β-glucopyranoside for screening studies
  • as a fluorogenic substrate to determine the glucocerebrosidase enzyme activity in substantia nigra

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Wen-Yih Jeng et al.
Journal of structural biology, 173(1), 46-56 (2010-08-05)
β-glucosidases (EC 3.2.1.21) cleave β-glucosidic linkages in disaccharide or glucose-substituted molecules and play important roles in fundamental biological processes. β-Glucosidases have been widely used in agricultural, biotechnological, industrial and medical applications. In this study, a high yield expression (70-250 mg/l)
Anna Lisa Gündner et al.
Neurobiology of disease, 121, 205-213 (2018-09-22)
Synucleinopathies including Parkinson's disease (PD) and Dementia with Lewy bodies (DLB) are characterized by the accumulation of abnormal α-synuclein in intraneuronal inclusions, named Lewy bodies. Mutations in GBA1, the gene encoding the lysosomal hydrolase glucocerebrosidase, have been identified as the
Pavel Mazura et al.
Carbohydrate research, 346(2), 238-242 (2010-12-15)
Here we present an optimized procedure to generate amino acid variations at specific site(s) of proteins, followed by a simple one-step screen for mutants with the desired β-glucosidase activity. The procedure was evaluated by introducing sequence variation into a codon
Wen-Yih Jeng et al.
Journal of structural biology, 173(1), 46-56 (2010-08-05)
β-glucosidases (EC 3.2.1.21) cleave β-glucosidic linkages in disaccharide or glucose-substituted molecules and play important roles in fundamental biological processes. β-Glucosidases have been widely used in agricultural, biotechnological, industrial and medical applications. In this study, a high yield expression (70-250 mg/l)

Articles

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Probiotics exhibit an inhibitory effect on pathogens, help prevent chronic intestinal inflammatory diseases or atopic syndromes, and support the immune system.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service