Skip to Content
Merck
All Photos(1)

Key Documents

Q2128

Sigma-Aldrich

Quisqualic acid

powder

Synonym(s):

β-(3,5-Dioxo-1,2,4-oxadiazolidin-2-yl)-L-alanine, 3-(3,5-Dioxo-1,2,4-oxadiazolidin-2-yl)-L-alanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H7N3O5
CAS Number:
Molecular Weight:
189.13
Beilstein:
1078734
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.32

form

powder

Quality Level

color

white to off-white

solubility

ethanol: <0.17 mg/mL
H2O: 0.5 mg/mL
0.1 M HCl: 1.4 mg/mL
1 M NH4OH: 20 mg/mL
organic solvents: insoluble

storage temp.

2-8°C

SMILES string

N[C@@H](CN1OC(=O)NC1=O)C(O)=O

InChI

1S/C5H7N3O5/c6-2(3(9)10)1-8-4(11)7-5(12)13-8/h2H,1,6H2,(H,9,10)(H,7,11,12)/t2-/m0/s1

InChI key

ASNFTDCKZKHJSW-REOHCLBHSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Quisqualate/ Quisqualic acid is obtained from the fruits and seeds of Quisqualis chinensis. It is an agonist at two subsets of excitatory amino acid receptors metabotropic receptors that indirectly mediate calcium mobilization from intracellular stores and, ionotropic receptors that directly control membrane channels. L-quisqualic acid is an agonist of the neurotransmitter L-glutamate.

Application

Quisqualic acid has also been used as a group I metabotropic receptor agonist in neurons.
Quisqualic acid has been used to test ligand-gated receptors in spiral ganglion neurons.

Biochem/physiol Actions

Active enantiomer of quisqualic acid; excitatory amino acid at glutamate receptors.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Optical recordings of Ca2+ signaling activities from identified inner ear cells in cochlear slices and hemicochleae
Xi Lin
Brain Res. Protoc., 11(2) (2003)
Sujeenthar Tharmalingam et al.
Neuropharmacology, 63(4), 667-674 (2012-06-02)
The metabotropic glutamate receptors (mGluRs) are evolutionarily conserved from nematodes to vertebrates. The Caenorhabditis elegans (C. elegans) genome contains three mGluR genes referred to as mgl-1, mgl-2, and mgl-3. The aim of this study was to characterize the pharmacological profiles
Pablo J Giraudi et al.
PloS one, 6(12), e29078-e29078 (2012-01-05)
We have previously reported that exposure of SH-SY5Y neuroblastoma cells to unconjugated bilirubin (UCB) resulted in a marked up-regulation of the mRNA encoding for the Na(+)-independent cystine∶glutamate exchanger System X(c)(-) (SLC7A11 and SLC3A2 genes). In this study we demonstrate that
Chinese Drugs of Plant Origin: Chemistry, Pharmacology, and Use in Traditional and Modern Medicine null
Thibault Collin et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 29(29), 9281-9291 (2009-07-25)
Little is known about the generation of slow rhythms in brain neuronal circuits. Nevertheless, a few studies, both from reconstituted systems and from hippocampal slices, indicate that activation of metabotropic glutamate receptors (mGluRs) could generate such rhythms. Here we show

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service