Skip to Content
Merck
All Photos(1)

Key Documents

PZ0370

Sigma-Aldrich

PD-85639

Synonym(s):

N-[3-(2,6-Dimethyl-1-piperidinyl)propyl]-α-phenylbenzeneacetamide, PD 85639, PD85,639, PD85639

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H32N2O
CAS Number:
Molecular Weight:
364.52
MDL number:
UNSPSC Code:
51111800
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (warmed)

storage temp.

room temp

SMILES string

CC1CCCC(C)N1CCCNC(C(C2=CC=CC=C2)C3=CC=CC=C3)=O

InChI

1S/C23H31N3O/c1-4-13-23(22(24)27,19-9-6-5-7-10-19)20-11-8-12-21(14-20)26-17(2)15-25-16-18(26)3/h5-12,14,17-18,25H,4,13,15-16H2,1-3H3,(H2,24,27)

InChI key

QNZVKSNHNMBPSR-UHFFFAOYSA-N

Biochem/physiol Actions

PD-85639 (PD85,639) is a voltage-gated sodium (Na+) channel blocker (75% in 10 min & >95% in 25 min blockage of Na+ current by 25 μM PD85,639; whole-cell patch clamp using primary rat brain neurons) that is shown to target rat brain Nav1.2 with simultaneous high- and low-affinity modes of binding (EC50 = 56 nM/40% & 20 μM/60% at pH 9.0, 5 nM/28% & 3 μM/72% at pH 7.4, against 2 nM [3H]-PD85,639 for binding rat brain synaptosomes; EC50 = 17 nM/39% & 10 μM/61% using at pH 9.0 using rat brain synaptosome membranes) and a fast kinetic (t1/2 = 1.2 at 4°C, <0.5 min at 25°C), competitive against the local anesthetic Na+ channel blockers tetracaine, bupivacaine, and mepivacaine, as well as Na+ channel activators veratridine and batrachotoxin (K1 = 0.26 μM against 5 nM [3H]-BTX for binding rat neocrotical membranes).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W Thomsen et al.
Molecular pharmacology, 43(6), 955-964 (1993-06-01)
The local anesthetic-like Na+ channel-blocking drug [3H]PD85639 [alpha-([4-3H]phenyl)-N-[3-(2,6-dimethyl-1-piperizinyl)-alpha-prop yl] [4-3H]benzeneacetamide] binds specifically to receptor sites on Na+ channels in intact synaptosomes and synaptosomal membranes, purified and reconstituted Na+ channels, and type IIA Na+ channel alpha subunits expressed in the transfected
I Roufos et al.
Journal of medicinal chemistry, 37(2), 268-274 (1994-01-21)
The synthesis and structure-activity relationships of a series of phenylacetamides related to N-[3-(2,6-dimethyl-1-piperidinyl)propyl]-alpha-phenylbenzeneacetamide (1) (PD85639) acting at the voltage-dependent Na+ channel are described. All structural variations for this study were made in the phenylacetic acid portion of these molecules, and
D S Ragsdale et al.
Molecular pharmacology, 43(6), 949-954 (1993-06-01)
This study examined the actions of the novel Na+ channel blocker PD85,639. In whole-cell voltage-clamp recordings from Chinese hamster ovary cells transfected with a cDNA encoding the rat brain type IIA Na+ channel and from dissociated rat brain neurons, PD85,639

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service