Skip to Content
Merck
All Photos(1)

Key Documents

PZ0320

Sigma-Aldrich

PF-04937319

≥98% (HPLC)

Synonym(s):

N, N-Dimethyl-5-((2-methyl-6-((5-methylpyrazin-2-yl)carbamoyl)benzofuran-4-yl)oxy)pyrimidine-2-carboxamide, N,N-Dimethyl-5-[[2-methyl-6-[[(5-methyl-2-pyrazinyl)amino]carbonyl]-4-benzofuranyl]oxy]-2-pyrimidinecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H20N6O4
CAS Number:
Molecular Weight:
432.43
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

CC(O1)=CC2=C1C=C(C(NC3=NC=C(C)N=C3)=O)C=C2OC4=CN=C(C(N(C)C)=O)N=C4

InChI

1S/C22H20N6O4/c1-12-8-24-19(11-23-12)27-21(29)14-6-17-16(5-13(2)31-17)18(7-14)32-15-9-25-20(26-10-15)22(30)28(3)4/h5-11H,1-4H3,(H,24,27,29)

InChI key

MASKQITXHVYVFL-UHFFFAOYSA-N

General description

PF-04937319 is a partial activator of glycokinase (GK) enzyme.

Biochem/physiol Actions

PF-04937319 is a glucokinase activator with an EC50 value of 174 nM. PF-04937319 was found to improve glycemic control in adults with type 2 diabetes when used in conjunction with metformin.
PF-04937319, unlike other glucokinase activators, is capable of maintaining lower-glucose levels without it resulting in hypoglycemia. In type II diabetic patients where metformin treatment is inadequate, PF-04937319 is capable of maintaining glycemic control within the acceptable risk-benefit profile.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

John C Pettersen et al.
Toxicologic pathology, 42(4), 696-708 (2014-04-29)
Glucokinase activators (GKAs) are being developed for the treatment of type 2 diabetes. The toxicity of 4 GKAs (PF-04279405, PF-04651887, piragliatin, and PF-04937319) was assessed in mice, rats, dogs, and/or monkeys. GKAs were administered for 2 to 8 weeks. Standard
N B Amin et al.
Diabetes, obesity & metabolism, 17(8), 751-759 (2015-04-18)
To assess the efficacy and safety of a range of doses of a systemic, partial, glucokinase activator, PF-04937319, as add-on therapy to metformin, in patients with type 2 diabetes mellitus (T2DM). Patients were randomized to once-daily PF-04937319 doses of 10
Raman Sharma et al.
Drug metabolism and disposition: the biological fate of chemicals, 42(11), 1926-1939 (2014-08-22)
The present article summarizes Metabolites in Safety Testing (MIST) studies on a glucokinase activator, N,N-dimethyl-5-((2-methyl-6-((5-methylpyrazin-2-yl)carbamoyl)benzofuran-4-yl)oxy)pyrimidine-2-carboxamide (PF-04937319), which is under development for the treatment of type 2 diametes mellitus. Metabolic profiling in rat, dog, and human hepatocytes revealed that PF-04937319 is

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service