Skip to Content
Merck
All Photos(1)

Key Documents

PZ0319

Sigma-Aldrich

PF-06409577

≥98% (HPLC)

Synonym(s):

6-Chloro-5-[4-(1-hydroxycyclobutyl)phenyl]-1H-indole-3-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H16ClNO3
CAS Number:
Molecular Weight:
341.79
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

room temp

SMILES string

ClC1=C(C2=CC=C(C3(O)CCC3)C=C2)C=C4C(NC=C4C(O)=O)=C1

InChI

1S/C19H16ClNO3/c20-16-9-17-14(15(10-21-17)18(22)23)8-13(16)11-2-4-12(5-3-11)19(24)6-1-7-19/h2-5,8-10,21,24H,1,6-7H2,(H,22,23)

InChI key

FHQXLWCFSUSXBF-UHFFFAOYSA-N

General description

PF-06409577 is a 6-chloro-indole derivative obtained from 5-bromo-6-chloro-indole.

Application

PF-06409577 has been used to determine its potential as a therapeutic agent for diabetic nephropathy.

Biochem/physiol Actions

PF-06409577 is a potent and selective activator of 5′ adenosine monophosphate-activated protein kinase (AMPK).
PF-06409577 potently activates a1β1γ1 AMPK (5′ adenosine monophosphate-activated protein kinase) isoform, and prevents its dephosphorylation. It is similarly potent for β1 containing isoforms, but shows significantly lower potency for β2-containing isoforms of AMPK. Patch-clamp assays show that this compound does not inhibit hERG (human ether-a-go-go gene). It interacts with the allosteric drug and metabolite site (ADaM) of AMPK.
PF-06409577 preserves retinal pigment epithelium cells from UV radiation by activating AMPK (AMP-activated protein kinase) signaling. This orally bioavailable and possesses pharmacokinetic properties. PF-06409577 blocks de novo lipid and cholesterol synthesis pathways that shows a decrease in hepatic lipids and mRNA expression of markers of hepatic fibrosis.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Activation of Liver AMPK with PF-06409577 Corrects NAFLD and Lowers Cholesterol in Rodent and Primate Preclinical Models.
Esquejo RM, et al.
EBioMedicine, 31, 122-132 (2018)
Nereida Jiménez de Oya et al.
Antimicrobial agents and chemotherapy, 62(7) (2018-05-02)
Mosquito-borne flaviviruses are a group of RNA viruses that constitute global threats for human and animal health. Replication of these pathogens is strictly dependent on cellular lipid metabolism. We have evaluated the effect of the pharmacological activation of AMP-activated protein
Discovery and Preclinical Characterization of 6-Chloro-5-[4-(1-hydroxycyclobutyl)phenyl]-1H-indole-3-carboxylic Acid (PF-06409577), a Direct Activator of Adenosine Monophosphate-activated Protein Kinase (AMPK), for the Potential Treatment of Diabetic Nephropathy.
Cameron KO et al
Journal of Medicinal Chemistry, 59(17), 8068-8081 (2016)
PF-06409577 activates AMPK signaling to protect retinal pigment epithelium cells from UV radiation.
Li X F, et al.
Biochemical and Biophysical Research Communications, 501(1), 293-299 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service