Skip to Content
Merck
All Photos(4)

Key Documents

PRS3141

Sigma-Aldrich

Anti-TLR4 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Toll-like receptor 4

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

predicted mol wt 92 kDa

species reactivity

human

technique(s)

immunocytochemistry: suitable
immunofluorescence: suitable
immunohistochemistry: suitable
indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TLR4(7099)

General description

Toll-like receptors (TLRs) are transmembrane receptors expressed on the surface of immune and non-immune cells. There are at least 13 different TLRs identified that mediate signalling pathways contributing to acute inflammatory processes during tissue damage and bacterial infections TLR2 is highly inducible by numerous inflammatory mediators resulting in translocation of transcription factor NF-κB-mediated proinflammatory cytokines and immune responses

Immunogen

a peptide corresponding to 15 amino acids near the amino-terminus of human TLR4.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3141.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amanda Croasdell et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 30(9), 3181-3193 (2016-06-04)
TLRs are critical for innate immunity, but excessive activation can lead to tissue damage and disease. Specialized proresolving mediators (SPMs), including resolvin D2 (RvD2), promote the active resolution of inflammation. How SPMs regulate early LPS signaling, including activation of TLR4
Shneh Sethi et al.
Virulence, 2(4), 316-328 (2011-07-16)
Recent studies in murine models of Staphylococcus aureus (S. aureus) infection have investigated the association of Toll-like receptor (TLR)2, TLR4, myeloid differentiation factor 88 (MyD88) and Nod-like receptor (NOD)2 with the production of cytokines / chemokines and their involvement in
Toll-like Receptor 4 Modulation as a Strategy to Treat Sepsis
Wittebole X et al
Mediators of Inflammation, doi: 10-doi: 10 (2010)
Emmanuel Lorne et al.
Intensive care medicine, 36(11), 1826-1835 (2010-08-07)
Although the role of Toll-like receptors (TLRs) in bacterial infection and sepsis is well characterized, recent studies have also shown that TLR4 and TLR2 can play an important role in contributing to acute inflammatory processes and organ dysfunction in settings

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service