Skip to Content
Merck
All Photos(1)

Key Documents

P9670

Sigma-Aldrich

Anti-Mouse IgG (whole molecule)–R-Phycoerythrin antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym(s):

GoatAnti-Mouse IgG (whole molecule)–R-PE

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

Quality Level

conjugate

phycoerythrin (R-PE) conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

form

buffered aqueous solution

technique(s)

indirect immunofluorescence: 1:20

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

General description

IgG is the main antibody type found in plasma and extracellular fluid and is expressed on B cell membrane. It plays a pivotal role in humoral immune responses like phagocytosis and complement activation.Anti-mouse IgG (whole molecule)-R-phycoerythrin antibody (diluted 1: 20) can be used as secondary antibody in fluorescence-activated cell sorting (FACS) analysis. It can also be used in flow cytometry. Goat anti-mouse IgG (whole molecule)-R-phycoerythrin antibody reacts specifically with mouse IgG and its subclasses IgG1, IgG2a, IgG2b, and IgG3. This product has also shown reactivity for mouse IgA and IgM but does not react with human serum proteins.

Immunogen

Purified mouse IgG.

Application

Anti-mouse IgG (whole molecule)-R-phycoerythrin antibody can be used as secondary antibody in immunocytofluorimetric analysis. It can also be used in immunohistochemistry and indirect immunofluorescence.

Other Notes

Antibody adsorbed with human serum proteins.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 0.1 mM EDTA, 1 mM iodoacetamide,
1% bovine serum albumin and 15 mM sodium azide.

Preparation Note

Adsorbed to reduce background with human samples.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Paul L Bollyky et al.
Journal of immunology (Baltimore, Md. : 1950), 179(2), 744-747 (2007-07-10)
Hyaluronan is a glycosaminoglycan present in the extracellular matrix. When hyaluronan is degraded during infection and injury, low m.w. forms are generated whose interactions influence inflammation and angiogenesis. Intact high m.w. hyaluronan, conversely, conveys anti-inflammatory signals. We demonstrate that high
Ira Bergman et al.
International journal of cancer, 121(2), 425-430 (2007-03-14)
Vesicular stomatitis virus (VSV) is being developed for cancer therapy. We have created a recombinant replicating VSV (rrVSV) that targeted to Her2/neu expressing breast cancer cells and expresses mouse GM-CSF. We now tested the efficacy of this rrVSV in the
Juliane Medler et al.
Cell death & disease, 10(3), 224-224 (2019-03-06)
Antibodies specific for TNFRSF receptors that bind soluble ligands without getting properly activated generally act as strong agonists upon FcγR binding. Systematic analyses revealed that the FcγR dependency of such antibodies to act as potent agonists is largely independent from
Michael Heyne et al.
Nature communications, 11(1), 297-297 (2020-01-17)
Quantifying the effects of various mutations on binding free energy is crucial for understanding the evolution of protein-protein interactions and would greatly facilitate protein engineering studies. Yet, measuring changes in binding free energy (ΔΔGbind) remains a tedious task that requires
C Pridgeon et al.
Rheumatology (Oxford, England), 42(7), 870-878 (2003-05-06)
Natural killer (NK) cells play an important role in several animal models of autoimmunity by modulating T-cell responses, but it is unclear whether human NK cells have similar functions. We characterized the phenotype of NK cells in synovial fluid (SF)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service