Skip to Content
Merck
All Photos(1)

Key Documents

P7265

Sigma-Aldrich

Prostaglandin A1

≥98% (HPLC), powder, NF-κB inhibitor

Synonym(s):

(13E,15S)-15-Hydroxy-9-oxoprosta-10,13-dien-1-oic acid, PGA1

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H32O4
CAS Number:
Molecular Weight:
336.47
Beilstein:
2003401
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.77

Product Name

Prostaglandin A1, synthetic

biological source

synthetic

Quality Level

Assay

≥98%

form

powder

storage temp.

−20°C

SMILES string

O=C1[C@H](CCCCCCC(O)=O)[C@@H](/C=C/[C@H](CCCCC)O)C=C1

InChI

1S/C20H32O4/c1-2-3-6-9-17(21)14-12-16-13-15-19(22)18(16)10-7-4-5-8-11-20(23)24/h12-18,21H,2-11H2,1H3,(H,23,24)/b14-12+/t16-,17-,18+/m0/s1

InChI key

BGKHCLZFGPIKKU-LDDQNKHRSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Biochem/physiol Actions

Prostaglandin A1 (PGA1) is a cyclopentenone prostaglandin containing chemically reactive α,β-unsaturated carbonyl. This cyclooxygenase 2 (COX-2) metabolic product aids in regulating the development and progression of Alzheimer′s disease (AD). In addition, PGA1 also regulates various biological processes via Michael addition. It exhibits antiviral activity against avian influenza A viruses (IAV) by inducing cytoprotective heat shock response in infected cells and by inhibiting nuclear factor-κB (NF-κB) activity.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guo-Biao Xu et al.
Molecular neurobiology, 58(3), 1114-1127 (2020-10-24)
Prostaglandin (PG) A1 is a metabolic product of cyclooxygenase 2 (COX-2) that is potentially involved in regulating the development and progression of Alzheimer's disease (AD). PGA1 is a cyclopentenone (cy) PG characterized by the presence of a chemically reactive α,β-unsaturated
A Aitokallio-Tallberg et al.
Cancer letters, 34(2), 201-206 (1987-02-01)
The production of the antiaggregatory and vasodilatory prostacyclin (PGI2) in patients with gynaecological tumours was studied by assaying urinary 6-keto-prostaglandin F1a (= 6-keto-PGF1a), a hydration product of PGI2), by radioimmunoassay following high performance liquid chromatography (HPLC) in 59 patients with
M G Santoro
Experientia, 50(11-12), 1039-1047 (1994-11-30)
Acute infection of mammalian cells with several types of RNA and DNA viruses often results in induction of heat-shock gene expression. The presence of hsp70 in intact virions, as well as the transient association of HSP with viral proteins and
Stefania Carta et al.
Prostaglandins, leukotrienes, and essential fatty acids, 91(6), 311-323 (2014-08-26)
Influenza A viruses (IAV) have the potential to cause devastating pandemics. In recent years, the emergence of new avian strains able to infect humans represents a serious threat to global human health. The increase in drug-resistant IAV strains underscores the
Beatriz Díez-Dacal et al.
Cancer research, 71(12), 4161-4171 (2011-04-22)
Cyclopentenone prostaglandins (cyPG) are reactive eicosanoids that may display anti-inflammatory and antiproliferative actions, possibly offering therapeutic potential. Here we report the identification of members of the aldo-keto reductase (AKR) family as selective targets of the cyPG prostaglandin A(1) (PGA(1)). AKR

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service