P6250
Methyl pentadecanoate
≥98.5%
Synonym(s):
Pentadecanoic acid methyl ester
About This Item
Recommended Products
Quality Level
Assay
≥98.5%
form
liquid
refractive index
n20/D 1.438 (lit.)
bp
141-142 °C/3 mmHg (lit.)
mp
18-19 °C (lit.)
density
0.862 g/mL at 25 °C (lit.)
functional group
ester
lipid type
saturated FAs
shipped in
ambient
storage temp.
room temp
SMILES string
CCCCCCCCCCCCCCC(=O)OC
InChI
1S/C16H32O2/c1-3-4-5-6-7-8-9-10-11-12-13-14-15-16(17)18-2/h3-15H2,1-2H3
InChI key
XIUXKAZJZFLLDQ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Protocols
Protocol for GC Analysis of Bacterial Acid Methyl Esters (BAMEs) on Equity®-1
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service