Skip to Content
Merck
All Photos(4)

Key Documents

P6125

Sigma-Aldrich

Pentadecanoic acid

~99% (capillary GC)

Synonym(s):

C15:0, NSC 28486, Pentadecylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13COOH
CAS Number:
Molecular Weight:
242.40
Beilstein:
1773831
EC Number:
MDL number:
UNSPSC Code:
12352211
PubChem Substance ID:
NACRES:
NA.25

biological source

synthetic (organic)

Quality Level

Assay

~99% (capillary GC)

form

powder or flakes

bp

257 °C/100 mmHg (lit.)

mp

51-53 °C (lit.)

functional group

carboxylic acid

lipid type

saturated FAs

shipped in

ambient

storage temp.

room temp

SMILES string

CCCCCCCCCCCCCCC(O)=O

InChI

1S/C15H30O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15(16)17/h2-14H2,1H3,(H,16,17)

InChI key

WQEPLUUGTLDZJY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jumat Salimon et al.
TheScientificWorldJournal, 2014, 906407-906407 (2014-04-11)
Two different procedures for the methylation of fatty acids (FAs) and trans fatty acids (TFAs) in food fats were compared using gas chromatography (GC-FID). The base-catalyzed followed by an acid-catalyzed method (KOCH3/HCl) and the base-catalyzed followed by (trimethylsilyl)diazomethane (TMS-DM) method
Yufan Zhang et al.
BMC plant biology, 14, 106-106 (2014-04-25)
The Arabidopsis thaliana LEC2 gene encodes a B3 domain transcription factor, which plays critical roles during both zygotic and somatic embryogenesis. LEC2 exerts significant impacts on determining embryogenic potential and various metabolic processes through a complicated genetic regulatory network. An
Ill-Min Chung et al.
Foods (Basel, Switzerland), 9(12) (2020-12-02)
Reliable geographical identification can protect producers of excellent agroproducts, and also provide reliable purchasing information to satisfy consumers. Therefore, this study aimed to evaluate the regional and monthly variation in fatty acid (FA) and tocopherol (TOC) in organic milk (OM)
Yufan Zhang et al.
Frontiers in plant science, 6, 239-239 (2015-05-01)
In plants, the conversion of stearoyl-ACP to oleoyol-ACP is catalyzed by a plastid-localized soluble stearoyl-acyl carrier protein (ACP) desaturase (SAD). The activity of SAD significantly impacts the ratio of saturated and unsaturated fatty acids, and is thus a major determinant
Ill-Min Chung et al.
Foods (Basel, Switzerland), 9(9) (2020-09-05)
Shiitake mushroom (Lentinula edodes) is commonly consumed worldwide and is cultivated in many farms in Korea using Chinese substrates owing to a lack of knowledge on how to prepare sawdust-based substrate blocks (bag cultivation). Consequently, issues related to the origin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service