Skip to Content
Merck
All Photos(1)

Key Documents

P5749

Sigma-Aldrich

S-(+)-PD 123177 trifluoroacetate salt hydrate

≥98% (HPLC), solid

Synonym(s):

(S)-1-[(4-Amino-3-methylphenyl)methyl]-5-(diphenylacetyl)-4,5,6,7-tetrahydro-1H-Imidazo[4,5-c]pyridine-6-carboxylic acid trifluoroacetate salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H28N4O3 · xC2HF3O2 · yH2O
Molecular Weight:
480.56 (anhydrous free base basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

solid

color

white to off-white

solubility

H2O: >5 mg/mL

storage temp.

2-8°C

SMILES string

O.OC(=O)C(F)(F)F.Cc1cc(Cn2cnc3CN([C@@H](Cc23)C(O)=O)C(=O)C(c4ccccc4)c5ccccc5)ccc1N

InChI

1S/C29H28N4O3.C2HF3O2.H2O/c1-19-14-20(12-13-23(19)30)16-32-18-31-24-17-33(26(29(35)36)15-25(24)32)28(34)27(21-8-4-2-5-9-21)22-10-6-3-7-11-22;3-2(4,5)1(6)7;/h2-14,18,26-27H,15-17,30H2,1H3,(H,35,36);(H,6,7);1H2/t26-;;/m0../s1

InChI key

IRRZMYRIDIBBQG-ROPHLPQBSA-N

Biochem/physiol Actions

S-(+)-PD 123177 is selective AT2 angiotensin receptor antagonist. The angiotensin AT2 receptor is an atypical seven transmembrane domain receptor that is coupled to activation of tyrosine phosphatase and inhibition of MAP kinase, and does not undergo agonist-induced internalization. An investigation of the occurrence and nature of AT2 receptor phosphorylation revealed that phorbol ester-induced activation of protein kinase C (PKC) in HA-AT2 receptor-expressing COS-7 cells caused rapid and specific phosphorylation of a single residue (Ser354) located in the cytoplasmic tail of the receptor. Agonist activation of AT2 receptors by angiotensin II (Ang II) also caused rapid PKC-dependent phosphorylation of Ser354 that was prevented by the AT2 antagonist, S-(+)-PD 123177, and by inhibitors of PKC. In cells coexpressing AT1 and AT2 receptors, Ang II-induced phosphorylation of the AT2 receptor was reduced by S-(+)-PD 123177 and abolished by treatment with both antagonists or with PKC inhibitors. These findings indicate that the AT2 receptor is rapidly phosphorylated via PKC during homologous activation by Ang II, and also undergoes heterologous PKC-dependent phosphorylation during activation of the AT1 receptor.

Features and Benefits

This compound is featured on the Angiotensin Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

We offer many products related to angiotensin receptors for your research needs.

We offer many products related to angiotensin receptors for your research needs.

We offer many products related to angiotensin receptors for your research needs.

We offer many products related to angiotensin receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service