Skip to Content
Merck
All Photos(3)

Key Documents

P5374

Sigma-Aldrich

Anti-Podoplanin antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Aggrus, Anti-GP36, Anti-Glycoprotein, 36-KD, Anti-OTS8, Anti-T1α gene, Anti-T1A, Anti-T1A2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~38 kDa

species reactivity

human, rat

concentration

~1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 10-20 μg/mL using heat retrieved, human skin sections
western blot (chemiluminescent): 5-10 μg/mL using human HEK-293 or rat NRK cells

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PDPN(10630)
rat ... Pdpn(54320)

General description

Podoplanin (PDPN) is a transmembrane sialoglycoprotein. It consists of 162 amino acids and is part of the type-1 transmembrane sialomucin-like glycoprotein family. The protein is expressed in various tissues. The PDPN gene is localized on human chromosome 1p36.21.
Podoplanin is an integral membrane mucoprotein present on the surface of podocytes in kidney glomeruli and the parietal cells of the glomerular Bowman′s capsule. It is a single membrane-spanning protein composed of a heavily glycosylated extracellular domain and a short intracytoplasmic tail. Podoplanin, also named E11 antigen, is present in normal, inflammatory and neoplastic lymphatic capillary endothelium. It is expressed selectively by dermal microvascular lymphatic endothelial cells but not by blood vascular endothelial cells.

Immunogen

synthetic peptide corresponding to amino acids 24-40 of human podoplanin with an N-terminal added cysteine, conjugated to KLH.

Application

Anti-Podoplanin antibody produced in rabbit has been used in immunoblotting.
Anti-Podoplanin antibody produced in rabbit has been used in immunohistochemistry.

Biochem/physiol Actions

Podoplanin (PDPN) is expressed in many cancers. In cancerous cells, it induces platelet aggregation and epithelial-mesenchymal transition. The protein is an immunohistochemical marker of lymphatic vessels. Podoplanin also has a role in embryonic development.
Podoplanin appears to play an important role in the maintenance of podocyte foot processes, and hence, glomerular permeability. It is detectable in human angiosarcomas, lymphangiomas and in Kaposi′s sarcomas. Experiments involving depletion of T1a/podoplanin in mice indicate a critical regulatory role for it in formation of the lymphatic vasculature.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide and 1% bovine serum albumin.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The multifaceted role of podoplanin expression in hepatocellular carcinoma.
Cioca A
European Journal of Histochemistry (2017)
Podoplanin associates with adverse postoperative prognosis of patients with clear cell renal cell carcinoma
Yu Xia
Cancer Science (2016)
Podoplanin Expression in Cancer-associated Fibroblasts Predicts Poor Prognosis in Patients with Squamous Cell Carcinoma of the Lung
Yohei Y
Anticancer Research (2017)
Activating PIK3CA alleles and lymphangiogenic phenotype of lymphatic endothelial cells isolated from lymphatic malformations
Alexander J. Osborn
Human Molecular Genetics (2015)
Abstract 877: Genetic variation in podoplanin in relation to glioma risk and outcome
Kristen N
Cancer Research (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service