Skip to Content
Merck
All Photos(1)

Key Documents

P5267

Sigma-Aldrich

L-Proline p-nitroanilide trifluoroacetate salt

≥99% (TLC), suitable for ligand binding assays

Synonym(s):

N-(4-Nitrophenyl)pyrrolidine-2-carboxamide, P-pNA, Pro-pNA

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13N3O3 · C2HF3O2
CAS Number:
Molecular Weight:
349.26
MDL number:
UNSPSC Code:
12352209
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.26

product name

L-Proline p-nitroanilide trifluoroacetate salt, prolyl aminopeptidase substrate

Quality Level

Assay

≥99% (TLC)

form

powder

technique(s)

ligand binding assay: suitable

color

white to yellow

storage temp.

2-8°C

SMILES string

OC(=O)C(F)(F)F.[O-][N+](=O)c1ccc(NC(=O)[C@@H]2CCCN2)cc1

InChI

1S/C11H13N3O3.C2HF3O2/c15-11(10-2-1-7-12-10)13-8-3-5-9(6-4-8)14(16)17;3-2(4,5)1(6)7/h3-6,10,12H,1-2,7H2,(H,13,15);(H,6,7)/t10-;/m0./s1

InChI key

KYRVEVYREUUAKH-PPHPATTJSA-N

General description

Proline p-nitroanilide (P-pNA) is a colorimetric substrate for prolyl aminopeptidase (proline iminopeptidase), an enzyme that releases proline from the N-terminus of small peptides.

Application

L-Proline p-nitroanilide trifluoroacetate salt has also been used as a monopeptide substrate for measuring the amidolytic activity of fibrillated peptide catalyst, PC4.
Proline p-nitroanilide (P-pNA) has been used as a substrate for prolyl aminopeptidase (proline iminopeptidase) from cabbage leaves.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yoke-Ming Wong et al.
Biomacromolecules, 17(10), 3375-3385 (2016-09-20)
Amyloid fibers are classified as a new generation of tunable bionanomaterials that exhibit new functions related to their distinctive characteristics, such as their universality, tunability, and stiffness. Here, we introduce the catalytic residues of serine protease into a peptide catalyst
Margarita Marinova et al.
Protein and peptide letters, 16(2), 207-212 (2009-02-10)
Chick-pea (Cicer arietinum L.) cotyledons are unique source of aminopeptidase - 8-9 U/g cotyledons was observed using L-leucine-p-nitroanilide as substrate. The aminopeptidase was purified (65 kDa, pI 4.8 ) reaching a specific activity of 220 U/mg at pH 7.0-7.2 and
Hongyu Yang et al.
World journal of microbiology & biotechnology, 32(11), 176-176 (2016-09-16)
Prolyl aminopeptidases are specific exopeptidases that catalyze the hydrolysis of the N-terminus proline residue of peptides and proteins. In the present study, the prolyl aminopeptidase gene (pap) from Aspergillus oryzae JN-412 was optimized through the codon usage of Pichia pastoris.
Paul J Lijnen et al.
Journal of the renin-angiotensin-aldosterone system : JRAAS, 6(2), 69-77 (2006-02-14)
To determine whether the aminopeptidase B inhibitor, arphamenine A, could affect collagen production and expression in control and TGF-ss1-treated cardiac fibroblasts. Cardiac fibroblasts from passage 2 from normal male adult rats were cultured to confluency and incubated with and without
Margarita Marinova et al.
Zeitschrift fur Naturforschung. C, Journal of biosciences, 63(1-2), 105-112 (2008-04-05)
Aminopeptidase, preferring phenylalanine-p-nitroanilide as substrate, and proline iminopeptidase, highly-specific for proline-p-nitroanilide, were isolated from cabbage leaves (Brassica oleraceae var. capitata). As pH optima, 7.2-7.5 for aminopeptidase activity and 8.0-8.5 for proline iminopeptidase were determined. Both peptidases were strongly inhibited by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service