Skip to Content
Merck
All Photos(2)

Key Documents

P4871

Sigma-Aldrich

Anti-Purinergic Receptor P2Y12 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym(s):

Anti-ADPG-R, Anti-BDPLT8, Anti-HORK3, Anti-P2T(AC), Anti-P2Y(12)R, Anti-P2Y(AC), Anti-P2Y(ADP), Anti-P2Y(cyc), Anti-P2Y12

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

lyophilized powder

species reactivity

human, mouse, rat

technique(s)

immunohistochemistry: suitable using mouse brain sections
western blot: 1:200 using rat brain membranes or human platelets

UniProt accession no.

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... P2RY12(64805)
mouse ... P2ry12(70839)
rat ... P2ry12(64803)

General description

Purinergic receptor or platelet adenosine 5′-diphosphate (ADP) receptor P2Y12 is a G-protein-coupled receptor. The gene encoding it is localized on human chromosome 3q25.1.

Immunogen

peptide corresponding to amino acid residues 125-142 of human P2Y12. This epitope is highly homologous in rat and mouse (16/18 residues identical).

Application

Anti-Purinergic Receptor P2Y12 antibody produced in rabbit has been used in Western blotting.

Biochem/physiol Actions

Purinergic receptor or platelet adenosine 5′-diphosphate (ADP) receptor P2Y12 has a role in platelet aggregation induced by ADP. Dysfunctional mutations in the protein have been linked with bleeding disorders. Studies have indicated that mice deficient in the P2Y12 receptor show decreased tumor metastasis.

Physical form

Lyophilized from phosphate buffered saline, pH 7.4, containing 1% bovine serum albumin and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 3

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inherited dysfunctional platelet P2Y12 receptor mutations associated with bleeding disorders.
Lecchi A
Hamostaseologie (2016)
Competitive mode and site of interaction of ticagrelor at the human platelet P2Y12-receptor
K. Hoffman
Journal of Thrombosis and Haemostasis (2014)
Leukotriene E4 induces MUC5AC release from human airway epithelial NCI-H292 cells
Hideaki Shirasaki
Allergology International (2014)
Supplementary Table 1. Sample demographics for participants included in genome-wide association analyses
Hypertension (2016)
Bernhard H Rauch et al.
Arteriosclerosis, thrombosis, and vascular biology, 30(12), 2434-2442 (2010-11-13)
The platelet P2Y12 ADP receptor is a well-known target of thienopyridine-type antiplatelet drugs. This study is the first to describe increased transcriptional expression of a functionally active P2Y12 in response to thrombin in human vascular smooth muscle cells (SMC). On

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service