P4404
Polyguanylic acid potassium salt
lyophilized powder
Synonym(s):
Poly(G) potassium salt
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
biological source
enzyme from (synthesis)
Quality Level
Assay
≥95% (TLC)
form
lyophilized powder
quality
≤5% free nucleotides
solubility
water: 19.60-20.40 mg/mL, clear to slightly hazy, colorless to light yellow
storage temp.
−20°C
Looking for similar products? Visit Product Comparison Guide
General description
Polyguanylic acid takes up a four-stranded helical structure. It is synthetically produced in Thermus thermophilus from guanosine diphosphate (GDP) and its synthesis is catalyzed by the enzyme polynucleotide phosphorylase.
Application
Polyguanylic acid (PolyG) is used to study the effects of ionizing radiation on formation and stability of G-quadruplex structures and as a target molecule for physicochemical studies of electronic excitation (vibrational spectra) of guanosine.
Polyguanylic acid potassium salt has been used:
- as a ligand for surface neuropilin-1 (NRP1) for internalization studies
- for intercalation studies with trisubstituted and disubstituted triazole-linked phenyl derivatives CL41, CL42 and CL2r50 using voltammetry method
- as a synthetic polynucleotide for agarose gel electrophoresis analysis
Biochem/physiol Actions
Polyguanylic acid acts as a ligand for scavenger receptor and reduces the surface expression neuropilin-1 in endothelial cells.
Preparation Note
Prepared from GDP using polynucleotide phosphorylase
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
The Journal of biological chemistry, 280(29), 26873-26879 (2005-05-27)
Little is currently known about the biochemical mechanism by which induced prion protein (PrP) conformational change occurs during mammalian prion propagation. In this study, we describe the reconstitution of PrPres amplification in vitro using partially purified and synthetic components. Overnight
Blood, 116(16), 3099-3107 (2010-07-08)
Ligand interaction with cognate cell-surface receptor often promotes receptor internalization, protecting cells from prolonged or excessive signaling from extracellular ligands. Compounds that induce internalization of surface receptors prevent ligand binding to cognate cell-surface receptors serving as inhibitors. Here, we show
Picosecond infrared probing of the vibrational spectra of transients formed upon UV excitation of stacked G-tetrad structures.
Chemical Communications (Cambridge, England), 28, 5158-5160 (2007)
Bioelectrochemistry (Amsterdam, Netherlands), 101, 97-105 (2014-09-10)
The redox mechanism of two trisubstituted triazole-linked phenyl derivatives (CL41 and CL42) and a disubstituted triazole-linked phenyl derivative (CL2r50) were studied using cyclic, differential pulse and square wave voltammetry at a glassy carbon electrode. The CL41, CL42 and CL2r50 oxidation
Radiation research, 173(1), 110-118 (2010-01-01)
The ability of guanine-rich sequences to form quadruplex structures in telomeres for example is important in a number of biological processes such as aging, carcinogenesis and gene regulation. Ionizing radiation can cause damage to guanine moieties that can affect the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service