Skip to Content
Merck
All Photos(1)

Key Documents

P2141

Sigma-Aldrich

Phalloidin Peptide

≥90% (HPLC), powder

Synonym(s):

28-(2,3-dihydroxy-2-methylpropyl)-18-hydroxy-34-(1-hydroxyethyl)-23,31-dimethyl-12-thia-10,16,22,25,27,30,33,36-octazapentacyclo[12.11.11.03,11.04,9.016,20]hexatriaconta-3(11),4,6,8-tetraene-15,21,24,26,29,32,35-heptone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H48N8O11S
CAS Number:
Molecular Weight:
788.87
Beilstein:
4347460
EC Number:
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.32

product name

Phalloidin from Amanita phalloides, ≥90%

biological source

Amanita phalloides

Quality Level

Assay

≥90%

form

powder

SMILES string

CC(O)C1NC(=O)C(C)NC(=O)C(CC(C)(O)CO)NC(=O)C2Cc3c(SCC(NC1=O)C(=O)N4CC(O)CC4C(=O)NC(C)C(=O)N2)[nH]c5ccccc35

InChI

1S/C35H48N8O11S/c1-15-27(47)38-22-10-20-19-7-5-6-8-21(19)41-33(20)55-13-24(34(53)43-12-18(46)9-25(43)31(51)37-15)40-32(52)26(17(3)45)42-28(48)16(2)36-30(50)23(39-29(22)49)11-35(4,54)14-44/h5-8,15-18,22-26,41,44-46,54H,9-14H2,1-4H3,(H,36,50)(H,37,51)(H,38,47)(H,39,49)(H,40,52)(H,42,48)

InChI key

KPKZJLCSROULON-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Phalloidin is a phallotoxin produced by death cap mushroom Amanita phalloides. It is a cyclic peptide, which interacts with actin, and this was first identified in phalloidin-poisoned rats. It is a heptapeptide, cyclic in nature, with a crosslink between tryptophan at position 6 and cysteine at position 3. The side chain of amino acid 7 (γ-δ-dihydroxyleucine) in phalloidin, is accessible to modifications, through which fluorescently labeled phalloidin compounds can be produced.

Application

Phalloidin has been used:
  • As a supplement in PEM buffer and dimethyl sulfoxide (DMSO).
  • As a drug.
  • In immunohistochemistry to stain F-actin.

Biochem/physiol Actions

Phalloidin interacts with polymeric actin, and not oligomeric or monomeric forms. This interaction leads to highly stabilized actin filaments, which resist depolymerization and disassembly. In rats, this toxin causes death due to liver hemorrhage and cells show abnormal actin clustering. The affinity of phalloidin to actin is not significantly altered after derivatizing fluorescently labelled phalloidin compounds. These compounds can be used to study actin structure and organization within eukaryotic cells.
Toxin that binds polymeric F actin, stabilizing it and interfering with the function of actin-rich structures.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thomas Pujol et al.
Proceedings of the National Academy of Sciences of the United States of America, 109(26), 10364-10369 (2012-06-13)
Actin filaments play a fundamental role in cell mechanics: assembled into networks by a large number of partners, they ensure cell integrity, deformability, and migration. Here we focus on the mechanics of the dense branched network found at the leading
Yaqi You et al.
Biomaterials, 269, 120356-120356 (2020-11-16)
Epicardial placement of mesenchymal stromal cells (MSCs) is a promising strategy for cardiac repair post-myocardial infarction, but requires the design of biomaterials to maximise the retention of donor cells on the heart surface and control their phenotype. To this end
Ilida Ortega et al.
Acta biomaterialia, 9(3), 5511-5520 (2012-11-08)
Corneal stem cell niches are located within the limbus of the eye and are believed to play an important role in corneal regeneration. These niches are often lost in corneal disease or trauma. Our work explores the design of artificial
Sheng Song et al.
Journal of neuroinflammation, 9, 219-219 (2012-09-20)
Activated microglial cells are an important pathological component in brains of patients with neurodegenerative diseases. The purpose of this study was to investigate the effect of He-Ne (632.8 nm, 64.6 mW/cm2) low-level laser therapy (LLLT), a non-damaging physical therapy, on activated microglia
Yueqin Wang et al.
PloS one, 8(9), e74051-e74051 (2013-09-17)
Hepatic stellate cell (HSC) plays a key role in pathogenesis of liver fibrosis. During liver injury, hypoxia in local micro-environment is inevitable. Hif-1α is the key transcriptional regulation factor that induces cell's adaptive responses to hypoxia. Recently, it was reported

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service