P1950
Pipet-Aid® automatic pipette pumps
model XP, cordless, rechargeable, AC/DC input 220 V AC, European 2-pin plug
All Photos(1)
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
Recommended Products
material
plastic
silicone
feature
CE compliant
manufacturer/tradename
Drummond Scientific 4-000-201
volume
1-100 mL
Related Categories
General description
- Pipet-Aid® automatic pipette is of light-weight and compact design.
- Three-speed range options (F) Fast, (M) Medium, (S) Slow provide more precise control for aspirating and dispensing samples.
- EZ Grip nosepiece with double membrane filter.
- Includes a removable stand.
- Easy access battery replacement.
- Holster-type wall bracket included.
Safe, convenient instruments for converting pipettes into hand-operated dispensers. Especially useful in confined areas such as laminar flow hoods. Can be used with glass or plastic pipettes ranging in size from 1 to 100 mL.
Legal Information
Pipet-Aid is a registered trademark of Drummond Scientific Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)