Skip to Content
Merck
All Photos(1)

Key Documents

P0071

Sigma-Aldrich

Anti-PDCD4 antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Neoplastic transformation inhibitor, Anti-Nuclear antigen H731, Anti-Programmed cell death 4, Anti-Protein 197/15a

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~51 kDa

species reactivity

human, mouse (predicted), rat (predicted)

concentration

~1 mg/mL

technique(s)

immunoprecipitation (IP): 5-10 μg using HEK-293T cell lysates
western blot: 2-4 μg/mL using K562 cell lysates

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PDCD4(272250)
mouse ... Pdcd4(18569)
rat ... SELP(6403)

General description

Programmed Cell Death 4 (PDCD4 ) is present in the nucleus. PDCD4 gene is mapped to human chromosome 10q25.2. PDCD4 protein consists of two basic domains at the N-terminus and C-terminus and two conserved α-helical MA-3 domains.

Specificity

Anti-PDCD4 specifically recognizes human, mouse, and rat PDCD4.

Application

Anti-PDCD4 antibody produced in rabbit has been used in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

Programmed Cell Death 4 (PDCD4) is a neoplastic transformation inhibitor protein that functions as a tumor suppressor in ovarian carcinoma. The levels of PDCD4 are depleted in progressed carcinomas of the lung, breast, colon, and prostate. MA-3 domain of PDCD4 aids protein-protein interactions in eukaryotic translation initiation factor 4GI(eIF4GI) and eIF4GII with the ATP-dependent RNA helicase eIF4A. PDCD4 suppresses translation initiation by specifically inhibiting the helicase activity of eukaryotic translation initiation factor 4A (eIF4A), a component of the translation initiation complex. In response to mitogens, PDCD4 is rapidly phosphorylated on Ser67 by the protein kinase ribosomal protein S6 kinase 1 (S6K1), and subsequently degraded via the ubiquitin ligase skp, cullin, f-box containing complex beta-transducin repeat-containing protein ((SCF)βTRCP ). It also shows that the protein is specifically phosphorylated on Ser76 and Ser457 by p90 ribosomal S6 kinase (RSK).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2–8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sachiko Matsuhashi et al.
International journal of molecular sciences, 20(9) (2019-05-12)
PDCD4 is a novel tumor suppressor to show multi-functions inhibiting cell growth, tumor invasion, metastasis, and inducing apoptosis. PDCD4 protein binds to the translation initiation factor eIF4A, some transcription factors, and many other factors and modulates the function of the
Brigitte Lankat-Buttgereit et al.
Biology of the cell, 101(6), 309-317 (2009-04-10)
Pdcd4 (programmed cell death 4) has been known as a tumour suppressor gene and potential target for anticancer therapies for several years. Initially, Pdcd4 was identified as a gene that is up-regulated during apoptosis, but its precise role still remains
Xialian Xu et al.
Kidney international, 82(11), 1167-1175 (2012-07-13)
Delayed ischemic preconditioning effectively protects kidneys from ischemia-reperfusion injury but the mechanism underlying renal protection remains poorly understood. Here we examined the in vivo role of microRNA miR-21 in the renal protection conferred by delayed ischemic preconditioning in mice. A
Sung-Chul Lim et al.
Oncology letters, 2(6), 1053-1057 (2012-10-11)
The aim of this study was to examine the role of Programmed cell death 4 (Pdcd4) in colorectal adenocarcinoma (CRA). Pdcd4 expression was observed in both the nucleus and cytoplasm in colorectal adenocarcinoma, whereas Pdcd4 was expressed in the nucleus
Jiali Long et al.
Gene, 680, 59-64 (2018-09-24)
In recent years, the incidence and mortality of colorectal cancer (CRC) have been on a global upward trend. There is an urgent need for effective tools to prevent and treat CRC and reduce morbidity and mortality of CRC patients. Recent

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service